Skip to Content
Merck
All Photos(1)

Key Documents

A1075

Sigma-Aldrich

Amyloid β Protein Fragment 1-40

≥90% (HPLC), powder

Synonym(s):

Aβ40

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C194H295N53O58S
CAS Number:
Molecular Weight:
4329.80
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.32

Quality Level

Assay

≥90% (HPLC)

form

powder

color

white

solubility

1% acetic acid: 1 mg/mL
saline: insoluble

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... APP(351)

Looking for similar products? Visit Product Comparison Guide

Amino Acid Sequence

Asp-Ala-Glu-Phe-Arg-His-Asp-Ser-Gly-Tyr-Glu-Val-His-His-Gln-Lys-Leu-Val-Phe-Phe-Ala-Glu-Asp-Val-Gly-Ser-Asn-Lys-Gly-Ala-Ile-Ile-Gly-Leu-Met-Val-Gly-Gly-Val-Val

General description

Amyloid β Protein Fragment 1-40 (Aβ40) is derived from the amyloid-β protein (Aβ), which is mapped to human chromosome 21q21.3. Aβ40 is predominantly present in the vascular amyloid deposits. Aβ40 comprises of C-terminal membrane insertion domain. It shows structural transition from random coil to a α-helical structure in a water-micelle medium.

Application

Amyloid β Protein Fragment 1-40 has been used:
  • in the temperature based conformational studies using Fourier transform infrared/differential scanning calorimetry (FT-IR/DSC) studies
  • as a reference standard in sandwich-type enzyme immunoassay for quantifying amyloid A4 protein in cerebrospinal fluid of patients with head trauma
  • as a component of embryonic stem cell medium to inhibit amyloid deposition in fibroblasts

Biochem/physiol Actions

Amyloid β Protein Fragment 1-40 (Aβ40) forms cation based ion channels.
Amyloid β-protein is neurotrophic and neurotoxic in vivo and in vitro in human and rat neuronal cell cultures. β-Amyloid peptides (amino acids 1-42 and 1-43) are the major constituents of senile plaques and neurofibrillary tangles that occur in the hippocampus, neocortex, and amygdala of patients with Alzheimer′s disease.

Reconstitution

For maximal biological activity, dilute the stock in calcium-free PBS to 1 mg/ml and incubate at 37 °C for 4 days.

Other Notes

Lyophilized from 0.1% TFA in H2O

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Amyloid beta protein 1-40 and 1-42 levels in matched cerebrospinal fluid and plasma from patients with Alzheimer disease
Mehta PD, et al.
Neuroscience Letters, 304(1-2), 102-106 (2001)
K Ueda et al.
Brain research, 639(2), 240-244 (1994-03-14)
The neurotoxic effects of soluble and aggregated synthetic amyloid beta protein (A beta P) have been investigated in rat primary cultures. Freshly solubilized beta(1-40) was neurotoxic not to immature, but to mature hippocampal neurons. On the other hand, aggregated beta(1-40)
Charlotte E Teunissen et al.
Journal of Alzheimer's disease : JAD, 62(4), 1857-1863 (2018-04-05)
The 42 amino acid form of amyloid-β (Aβ42) plays a key role in the pathogenesis of Alzheimer's disease (AD) and is a core biomarker for the diagnosis of AD. Numerous studies have shown that cerebrospinal fluid (CSF) Aβ42 concentrations are
Temperature-induced conformational changes in amyloid beta (1-40) peptide investigated by simultaneous FT-IR microspectroscopy with thermal system
Chu HL and Lin SY
Biophysical Chemistry, 89(2-3), 173-180 (2001)
Sandwich-Type Enzyme Immunoassay for Amyloid A4 Protein in Cerebrospinal Fluid From Patients with Head Trauma
PIRIM I
Turkish Journal of Medical Sciences, 31(1), 47-50 (2001)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service