Skip to Content
Merck
All Photos(4)

Documents

A0937

Sigma-Aldrich

(±)-Norepinephrine (+)-bitartrate salt

≥97% (TLC), powder, adrenergic neurotransmitter

Synonym(s):

(±)-Arterenol (+)-bitartrate salt, (±)-Noradrenalin (+)-bitartrate salt, 1-(3,4-Dihydroxyphenyl)-2-aminoethanol (+)-bitartrate salt, 3,4-Dihydroxyphenylethanolamine (+)-bitartrate salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H11NO3 · C4H6O6
CAS Number:
Molecular Weight:
319.26
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

product name

(±)-Norepinephrine (+)-bitartrate salt,

form

powder

Quality Level

SMILES string

C[C@H]([C@@H](O)C(O)=O)C(O)=O.NCC(O)c1ccc(O)c(O)c1

InChI

1S/C8H11NO3.C5H8O5/c9-4-8(12)5-1-2-6(10)7(11)3-5;1-2(4(7)8)3(6)5(9)10/h1-3,8,10-12H,4,9H2;2-3,6H,1H3,(H,7,8)(H,9,10)/t;2-,3-/m.1/s1

InChI key

PDZZSMDCBWCGRN-GEHKUQKJSA-N

Biochem/physiol Actions

Adrenergic neurotransmitter

Features and Benefits

This compound is featured on the α1-Adrenoceptors, α2-Adrenoceptors and β-Adrenoceptors pages of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 2 Dermal - Acute Tox. 2 Oral

Storage Class Code

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stéphanie Falcao et al.
American journal of physiology. Heart and circulatory physiology, 296(1), H119-H126 (2008-11-04)
The cyclin-dependent kinase inhibitor p57(kip2) regulates the cell cycle of trophoblastic cells. It has been established by a Japanese group that the heterozygous p57(kip2) knockout (p57(-/+)) mice are a good model of preeclampsia as they develop hypertension, proteinuria, and placental
[Hypovolemic shock during pregnancy].
E M Shifman
Anesteziologiia i reanimatologiia, (6)(6), 63-66 (2013-05-15)
Guoqing Di et al.
Noise & health, 15(65), 217-223 (2013-06-19)
Studies have reported that railway noise causes stress responses. To evaluate the effects of high-speed railway (HSR) noise on behaviors and plasma monoamines. Institute of cancer research mice were exposed to previously recorded HSR noise for 53 days. The noise
[Tapentadol prolonged release for severe chronic pain. Results of a non-interventional study involving general practitioners and internists].
Andreas Schwittay
MMW Fortschritte der Medizin, 155(1), 64-64 (2013-04-12)
Takahiro Higuchi et al.
Journal of nuclear medicine : official publication, Society of Nuclear Medicine, 54(7), 1142-1146 (2013-05-15)
A novel (18)F-labeled tracer, LMI1195 (N-[3-bromo-4-(3-(18)F-fluoro-propoxy)-benzyl]-guanidine), is being developed for sympathetic nerve imaging; its high specificity for neural uptake-1 mechanism has previously been demonstrated in cell associative studies and in rabbit and nonhuman primate studies assessing heart uptake. The aim

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service