Skip to Content
Merck
All Photos(1)

Documents

A0855

Sigma-Aldrich

Anti-Auxin antibody, Mouse monoclonal

clone 1E11-C11, purified from hybridoma cell culture

Synonym(s):

Anti-IAA, Anti-Indole-3-acetic acid

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

recombinant

expressed in mouse cell line

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

1E11-C11, monoclonal

form

PBS solution

species reactivity

plant

technique(s)

dot blot: suitable
immunofluorescence: suitable
immunohistochemistry: suitable
indirect ELISA: 1-2 μg/mL using using IAA conjugated to BSA
western blot: suitable

isotype

IgG2b

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

General description

Auxin is a plant hormone that regulates physiological growth and development. This hormone regulates cellular functions such as elongation, turgor, division, and differentiation. Indoleacetic acid (IAA) is recognized as the main auxin in several plants. IAA is synthesized from tryptophan (Trp) using Trp dependent pathways,. Mouse monoclonal anti-auxin antibody recognizes indoleacetic acid but not free unmethylated IAA.

Immunogen

indoleacetic acid (IAA) carboxyl linked to ovalbumin.

Application

Indole-3-acetic acid was identified in developing P.abies spruce plant parrafin embedded formalin-fixed embryos by immunohistochemistry with the monoclonal anti-indole-3-acetic acid antibody.
Mouse monoclonal anti-auxin antibody has been used for immunofluorescence studies. The antibody can also be used for ELISA, dot blot and western blot analyses.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T Mori et al.
American journal of reproductive immunology (New York, N.Y. : 1989), 26(3), 97-103 (1991-10-01)
Expression of CD4-like molecule on vitelline membrane of murine eggs was demonstrated by indirect immunofluorescence (IIF) test and immunoprecipitation corresponding to the expression of major histocompatibility complex (MHC) class II molecule on murine sperm detected by immunoblotting. This molecule showed
Yue Liang et al.
The Plant cell, 32(11), 3485-3499 (2020-08-28)
Developmental transitions in plants require adequate carbon resources, and organ abscission often occurs due to competition for carbohydrates/assimilates. Physiological studies have indicated that organ abscission may be activated by Suc deprivation; however, an underlying regulatory mechanism that links Suc transport
Manjul Singh et al.
Scientific reports, 7(1), 16101-16101 (2017-11-25)
Plants possess exuberant plasticity that facilitates its ability to adapt and survive under challenging environmental conditions. The developmental plasticity largely depends upon cellular elongation which is governed by a complex network of environmental and phytohormonal signals. Here, we report role
Shuai Zheng et al.
Plant communications, 2(5), 100134-100134 (2021-11-09)
In trees, stem secondary growth depends on vascular cambium proliferation activity and subsequent cell differentiation, in which an auxin concentration gradient across the cambium area plays a crucial role in regulating the process. However, the underlying molecular mechanism for the
Tadashi Sakata et al.
Proceedings of the National Academy of Sciences of the United States of America, 107(19), 8569-8574 (2010-04-28)
With global warming, plant high temperature injury is becoming an increasingly serious problem. In wheat, barley, and various other commercially important crops, the early phase of anther development is especially susceptible to high temperatures. Activation of auxin biosynthesis with increased

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service