Skip to Content
Merck
All Photos(1)

Key Documents

A0414

Sigma-Aldrich

p-Aminobenzyl 1-thio-β-D-galactopyranoside–Agarose

saline suspension

Synonym(s):

β-Galactopyranoside Agarose, Agarose for Galactopyranoside, Aminobenzyl Agarose

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12161501
NACRES:
NA.32

form

saline suspension

matrix

cross-linked 4% beaded agarose

matrix activation

cyanogen bromide

matrix attachment

amino

matrix spacer

12 atoms

capacity

≥10 mg/mL, gel binding capacity (β-galactosidase)

storage temp.

2-8°C

SMILES string

[X]Nc1ccc(cc1)CS[C@@H]2O[C@@H]([C@@H]([C@@H]([C@H]2O)O)O)CO

Application

p-Aminobenzyl 1-thio-β-D-galactopyranoside−Agarose has been used in the column for the purification of Bacillus circulans sp. alkalophilus b-galactosidase (Bca-b-gal). It has also been used in the derivatization of affinity matrix of agarose to perform affinity chromatography for the purification of P. haloplanktis β-galactosidase.

Physical form

Suspension in 0.5 M NaCl containing preservative

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Structural analysis, enzymatic characterization, and catalytic mechanisms of beta-galactosidase from Bacillus circulans sp. alkalophilus
Maksimainen M, et al.
The FEBS journal, 279(10), 1788-1798 (2012)
Cold-adapted beta-galactosidase from the Antarctic psychrophile Pseudoalteromonas haloplanktis
Hoyoux A, et al.
Applied and Environmental Microbiology, 67(4), 1529-1535 (2001)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service