Skip to Content
Merck
All Photos(1)

Key Documents

A0144

Sigma-Aldrich

Adenosine 3′,5′-cyclic monophosphate–Agarose

lyophilized powder

Synonym(s):

3′,5′-cAMP agarose

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
41106500
eCl@ss:
32160414
NACRES:
NA.56

form

lyophilized powder

extent of labeling

≥2 μmol per mL

matrix

4% beaded agarose

matrix activation

cyanogen bromide

matrix attachment

C-8

matrix spacer

9 atoms

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Application

Adenosine 3′,5′-cyclic monophosphate-Agarose (3′,5′-cAMP agarose) may be used in affinity chromatography for the separation of the subunits of cAMP-dependent protein kinase or for the purification of the cAMP receptor subunit. Research on schizophrenia has shown that abnormalities in the cAMP signaling pathway, may contribute to the pathophysiology of the disorder.

Physical form

Lyophilized powder stabilized with lactose

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Meyer et al.
Journal of bacteriology, 183(18), 5248-5256 (2001-08-22)
Klebsiella pneumoniae is able to grow anaerobically with citrate as a sole carbon and energy source by a fermentative pathway involving the Na(+)-dependent citrate carrier CitS, citrate lyase, and oxaloacetate decarboxylase. The corresponding genes are organized in the divergent citC
R G Panchal et al.
The Journal of biological chemistry, 269(38), 23722-23730 (1994-09-23)
cAMP-dependent protein kinase (PKA) is an important participant in neuronal modulation: the ability of neurons to change their properties in response to external stimuli. In Aplysia mechanosensory neurons, PKA plays roles in both short and long term presynaptic facilitation, which
K Laukens et al.
FEBS letters, 508(1), 75-79 (2001-11-15)
The soluble protein fraction of tobacco bright yellow 2 cells contained adenosine 3',5'-cyclic monophosphate (cAMP)-binding activity, detected with both a conventional binding assay and a surface plasmon resonance biosensor. A cAMP-agarose-based affinity purification procedure yielded three proteins which were identified
Adam J Funk et al.
Neuropsychopharmacology : official publication of the American College of Neuropsychopharmacology, 37(4), 896-905 (2011-11-04)
Recent evidence suggests that schizophrenia may result from alterations of integration of signaling mediated by multiple neurotransmitter systems. Abnormalities of associated intracellular signaling pathways may contribute to the pathophysiology of schizophrenia. Proteins and phospho-proteins comprising mitogen activated protein kinase (MAPK)
Ganesh Anand et al.
Biochemistry, 46(32), 9283-9291 (2007-07-31)
To better understand the molecular mechanism of cAMP-induced and substrate-enhanced activation of type-I A-kinase, we measured the kinetics of A-kinase regulatory subunit interactions using a stopped-flow spectrofluorometric method. Specifically, we conjugated fluorescein maleimide (FM) to two separate single cysteine-substituted and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service