Skip to Content
Merck
All Photos(1)

Key Documents

96669

Sigma-Aldrich

7-Diethylamino-3-(4-maleimidophenyl)-4-methylcoumarin

suitable for fluorescence, BioReagent, ≥90% (HPCE)

Synonym(s):

1H-Pyrrole-2,5-dione, 1-(4-(7-(diethylamino)-4-methyl-2-oxo-2H-1-benzopyran-3-yl)phenyl)-, CPM

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H22N2O4
CAS Number:
Molecular Weight:
402.44
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.32

product line

BioReagent

Assay

≥90% (HPCE)

form

solid

fluorescence

λex 391 nm; λem 472 nm in methanol: 2-mercaptoethanol

suitability

suitable for fluorescence

storage temp.

−20°C

SMILES string

CCN(CC)c1ccc2C(C)=C(C(=O)Oc2c1)c3ccc(cc3)N4C(=O)C=CC4=O

InChI

1S/C24H22N2O4/c1-4-25(5-2)18-10-11-19-15(3)23(24(29)30-20(19)14-18)16-6-8-17(9-7-16)26-21(27)12-13-22(26)28/h6-14H,4-5H2,1-3H3

InChI key

YGIABALXNBVHBX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Fluorescence probe for glutathione for intact cells; specific advantages: only thio derivative fluoresces, high quantum yield and good Stokes shift

Biochem/physiol Actions

Used to monitor release of thiols, quantitate thiol in microplate reactions, and to distinguish proliferating cancer cells by nucleolar protein staining.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Lutsenko et al.
Biochemistry, 32(26), 6737-6743 (1993-07-06)
The role of the Na,K-ATPase beta-subunit in stabilization of ion-binding sites has been investigated. Treatment of the purified renal Na,K-ATPase with 0.25 M DTT at 40 degrees C for 1 h resulted in 50% loss of Rb occlusion, which correlates
Simple Futarmal Kothari et al.
Pain, 156(12), 2545-2555 (2015-08-27)
The pathophysiology and underlying pain mechanisms of temporomandibular disorders (TMD) are poorly understood. The aims were to assess somatosensory function at the temporomandibular joints (TMJs) and to examine whether conditioned pain modulation (CPM) differs between TMD pain patients (n =
Evys Collazo et al.
Analytical biochemistry, 342(1), 86-92 (2005-06-17)
Histone methyltransferases (HMTs) catalyze the S-adenosylmethionine (AdoMet)-dependent methylation of lysines and arginines in the nucleosomal core histones H3 and H4 and the linker histone H1b. Methylation of these residues regulates either transcriptional activation or silencing, depending on the residue modified
Andrew A Voss et al.
The Journal of biological chemistry, 279(33), 34514-34520 (2004-06-16)
The skeletal-type ryanodine receptor (RyR1) undergoes covalent adduction by nitric oxide (NO), redox-induced shifts in cation regulation, and non-covalent interactions driven by the transmembrane redox potential that enable redox sensing. Tight redox regulation of RyR1 is thought to be primarily
G Liu et al.
Molecular pharmacology, 45(2), 189-200 (1994-02-01)
The fluorogenic sulfhydryl probe 7-diethylamino-3-(4'-maleimidylphenyl)-4-methylcoumarin (CPM) (1-50 nM) is used to characterize the functional role and location of highly reactive thiol groups on the ryanodine-sensitive Ca2+ release channel complex [i.e., ryanodine receptors (RyRs)] of skeletal and cardiac junctional sarcoplasmic reticulum

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service