94968
Polysucrose 40-fluorescein isothiocyanate conjugate
average mol wt 30,000-50,000
Synonym(s):
FITC-Polysucrose 40, Fluorescein isothiocyanate-Polysucrose 40 conjugate
About This Item
Recommended Products
form
powder
mol wt
average mol wt 30,000-50,000
fluorescence
λex 498 nm; λem 521 nm in 0.1 M phosphate pH 7.0
Analysis Note
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
Explore FITC-labelled polysaccharides and their properties, including impact of molecular weight on molecule dimensions.
Explore FITC-labelled polysaccharides and their properties, including impact of molecular weight on molecule dimensions.
Explore FITC-labelled polysaccharides and their properties, including impact of molecular weight on molecule dimensions.
Explore FITC-labelled polysaccharides and their properties, including impact of molecular weight on molecule dimensions.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service