Skip to Content
Merck
All Photos(1)

Key Documents

94370

Sigma-Aldrich

Uridine 5′-triphosphate trisodium salt dihydrate

≥80%

Synonym(s):

UTP

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C9H12N2O15P3Na3
CAS Number:
Molecular Weight:
586.12
Beilstein:
3585234
EC Number:
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.51
Pricing and availability is not currently available.

Quality Level

Assay

≥80%

form

powder

impurities

≤15% UDP
≤5% UMP

mp

>140 °C (dec.) (lit.)

shipped in

wet ice

storage temp.

−20°C

SMILES string

[Na+].[Na+].[Na+].[H]O[H].[H]O[H].O[C@H]1[C@@H](O)[C@@H](O[C@@H]1COP([O-])(=O)OP([O-])(=O)OP(O)([O-])=O)N2C=CC(=O)NC2=O

InChI

1S/C9H15N2O15P3.3Na.2H2O/c12-5-1-2-11(9(15)10-5)8-7(14)6(13)4(24-8)3-23-28(19,20)26-29(21,22)25-27(16,17)18;;;;;/h1-2,4,6-8,13-14H,3H2,(H,19,20)(H,21,22)(H,10,12,15)(H2,16,17,18);;;;2*1H2/q;3*+1;;/p-3/t4-,6-,7-,8-;;;;;/m1...../s1

InChI key

KIYVKLDQLQZXKH-GTIFRNKBSA-K

Gene Information

human ... P2RY2(5029)
rat ... P2ry2(29597)

Looking for similar products? Visit Product Comparison Guide

Application

Uridine 5′-diphosphate (UDP) and Uridine 5′-triphosphate (UTP) may be used in studies on nucleic acid (RNA) biosynthesis and cell signaling. UDP is a nucleotide that upon phosphorylation to UTP becomes a substrate for enzymes such as RNA polymerase(s) and GTPases. These enzymes are involved in a wide range of applications from the synthesis of RNA to the regulation of G-coupled Protein Receptors (GPCR) and cell signaling molecules such as Rho-signaling via Guanine Nucleotide Exchange Factors (GEF).

Biochem/physiol Actions

P2Y receptor agonist.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Feng Ding et al.
Current topics in medicinal chemistry, 11(23), 2879-2887 (2011-08-10)
The Rho family small GTPases of the Ras superfamily play key roles in regulating diverse signaling pathways that control a myriad of fundamental cellular processes such as cytoskeletal dynamics, cell cycle progression, gene expression, cell polarity, migration and cell transformation.
Caroline Liot et al.
PloS one, 6(8), e23676-e23676 (2011-09-03)
Besides regulation of actin cytoskeleton-dependent functions, Rho GTPase pathways are essential to cell cycle progression and cell division. Rho, Rac and Cdc42 regulate G1 to S phase progression and are involved in cytokinesis. RhoA GDP/GTP cycling is required for normal
Markie O Muoboghare et al.
British journal of pharmacology, 176(16), 2894-2904 (2019-05-23)
There is a lack of potent, selective antagonists at most subtypes of P2Y receptor. The aims of this study were to characterise the pharmacological properties of the proposed P2Y2 receptor antagonist, AR-C118925XX, and then to use it to determine the
Sara J Wright et al.
Genetics, 189(1), 165-176 (2011-07-14)
Heterotrimeric (αβγ) G proteins are crucial components of eukaryotic signal transduction pathways. G-protein-coupled receptors (GPCRs) act as guanine nucleotide exchange factors (GEFs) for Gα subunits. Recently, facilitated GDP/GTP exchange by non-GPCR GEFs, such as RIC8, has emerged as an important
Jae-Hyoung Song et al.
Antiviral research, 145, 6-13 (2017-07-15)
Rhinovirus, a major causative agent of the common cold, is associated with exacerbation of asthma and chronic obstructive pulmonary disease. Currently, there is no antiviral treatment or vaccine for human rhinovirus (HRV). Gemcitabine (2',2'-difluorodeoxycytidine, dFdC) is a deoxycytidine analog with

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service