Skip to Content
Merck

Skip To

92001

Ampliflu Red "easy to use" Kit

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

NACRES:
NA.32
UNSPSC Code:
12352108
Storage temp.:
2-8°C
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

powder

Quality Segment

fluorescence

λex 571 nm; λem 585 nm

storage temp.

2-8°C

General description

For the easy preparation of Ampliflu Red peroxidase substrate working solutions. Includes preweighed vials of Ampliflu Red and solvent (DMSO).

Legal Information

Ampliflu is a trademark of Sigma-Aldrich Co. LLC

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
90101AMP31004465962
fluorescence

λex 571 nm; λem 585 nm

fluorescence

λex 571 nm; λem 585 nm in DMSO

fluorescence

-

fluorescence

-

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

-

storage temp.

−20°C

form

powder

form

powder

form

-

form

-

Quality Level

100

Quality Level

100

Quality Level

-

Quality Level

-


Kit Components Only

Product No.
Description

  • Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC) 5 mL

Kit Components Also Available Separately

Product No.
Description
SDS & Pricing

  • Ampliflu Red, suitable for fluorescence, ≥98.0% (HPLC) 10 x 1
    SDS

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters