Skip to Content
Merck
All Photos(1)

Key Documents

78555

Sigma-Aldrich

Phenyl-β-D-glucuronide

≥99.0% (HPLC), powder

Synonym(s):

Phenyl-β-D-glucopyranosiduronic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H14O7
CAS Number:
Molecular Weight:
270.24
Beilstein:
89117
EC Number:
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.83

product name

Phenyl-β-D-glucuronide, ≥99.0% (HPLC)

Assay

≥99.0% (HPLC)

form

powder

optical activity

[α]20/D −87±5°, c = 1% in H2O

impurities

≤8% water

storage temp.

2-8°C

SMILES string

O[C@@H]1[C@@H](O)[C@@H](O[C@@H]([C@H]1O)C(O)=O)Oc2ccccc2

InChI

1S/C12H14O7/c13-7-8(14)10(11(16)17)19-12(9(7)15)18-6-4-2-1-3-5-6/h1-5,7-10,12-15H,(H,16,17)/t7-,8-,9+,10-,12+/m0/s1

InChI key

WVHAUDNUGBNUDZ-GOVZDWNOSA-N

General description

Phenyl-β-D-glucuronide is majorly used as a reference standard in chromatographic and mass spectrophotometry studies for quantifying glucuroconjugate metabolites in plasma and urine samples.

Application

Phenyl-β-D-glucuronide has been used as a nonpeptide chromogen for testing biuret reagent and as a reference standard for phenol analysis in urine samples.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Development of a class-selective enzyme immunoassay for urinary phenolic glucuronides
Staimer N, et al.
Analytica Chimica Acta, 444(1), 27-36 (2001)
Toluene, ethylbenzene and phenol-Determination of o-cresol, m-cresol, 2-ethylphenol, 4-ethylphenol and phenol in urine using gas chromatography-mass spectrometry [Biomonitoring Methods, 2011]
Schettgen T, et al.
The MAK-Collection for Occupational Health and Safety, 1-31 (2002)
Quantitative measurement of propofol and in main glucuroconjugate metabolites in human plasma using solid phase extraction-liquid chromatography-tandem mass spectrometry
Cohen S, et al.
Journal of Chromatography. B, Biomedical Sciences and Applications, 854(1-2), 165-172 (2007)
Quantitative amino acid and proteomic analysis: very low excretion of polypeptides> 750 Da in normal urine
Norden AGW, et al.
Kidney International, 66(5), 1994-2003 (2004)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service