Skip to Content
Merck
All Photos(1)

Key Documents

76508

Sigma-Aldrich

Atto 647N Protein Labeling Kit

BioReagent, suitable for fluorescence

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352108
NACRES:
NA.32

product line

BioReagent

manufacturer/tradename

ATTO-TEC GmbH

fluorescence

λex 647 nm; λem 661 nm in 0.1 M phosphate buffer, pH 7.0 (recommended)

suitability

suitable for fluorescence

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

This kit contains sufficient amounts of reactive dye, buffers and protein purification sets for performing five labeling reactions (1 mg protein each) and for the subsequent purification of the labeled protein.

Legal Information

This product is for Research use only. In case of intended commercialization, please contact the IP-holder (ATTO-TEC GmbH, Germany) for licensing.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bryant L Doss et al.
Proceedings of the National Academy of Sciences of the United States of America, 117(23), 12817-12825 (2020-05-24)
Morphogenesis, tumor formation, and wound healing are regulated by tissue rigidity. Focal adhesion behavior is locally regulated by stiffness; however, how cells globally adapt, detect, and respond to rigidity remains unknown. Here, we studied the interplay between the rheological properties
Anne Chalumeau et al.
Bio-protocol, 11(2), e3899-e3899 (2021-03-19)
β-hemoglobinopathies are severe genetic disorders characterized either by the abnormal synthesis of the adult β-globin chains of the hemoglobin (Hb) tetramer (βS-globin chains) in sickle cell disease (SCD) or by the reduced β-globin production in β-thalassemia. The identification and quantification
Alexandra Grubman et al.
Nature communications, 12(1), 3015-3015 (2021-05-23)
The role of microglia cells in Alzheimer's disease (AD) is well recognized, however their molecular and functional diversity remain unclear. Here, we isolated amyloid plaque-containing (using labelling with methoxy-XO4, XO4+) and non-containing (XO4-) microglia from an AD mouse model. Transcriptomics
Shiqiong Hu et al.
Molecular biology of the cell, 30(16), 2025-2036 (2019-06-20)
Contractile arrays of actin and myosin II filaments drive many essential processes in nonmuscle cells, including migration and adhesion. Sequential organization of actin and myosin along one dimension is followed by expansion into a two-dimensional network of parallel actomyosin fibers
Analysis of replication factories in human cells by super-resolution light microscopy.
Cseresnyes, Z.; et al.
BMC Cell Biology, 10(1), 88-88 (2009)

Articles

Protein labeling kits with Atto and Tracy dyes provide easy fluorescent labeling of purified proteins, enzymes, and antibodies.

Protein labeling kits with Atto and Tracy dyes provide easy fluorescent labeling of purified proteins, enzymes, and antibodies.

Protein labeling kits with Atto and Tracy dyes provide easy fluorescent labeling of purified proteins, enzymes, and antibodies.

Protein labeling kits with Atto and Tracy dyes provide easy fluorescent labeling of purified proteins, enzymes, and antibodies.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service