Skip to Content
Merck
All Photos(1)

Key Documents

72464

Supelco

Atto 565 NHS ester

BioReagent, suitable for fluorescence, ≥90% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C35H34ClN3O11
Molecular Weight:
708.11
MDL number:
UNSPSC Code:
12352108
NACRES:
NA.32

product line

BioReagent

Assay

≥90% (HPLC)
≥90% (degree of coupling)

form

powder

manufacturer/tradename

ATTO-TEC GmbH

suitability

suitable for fluorescence

detection method

fluorometric

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

Atto 565 NHS ester is a commonly used amine-reactive reagent, provided by ATTO-TEC. It is widely used in protein labeling and readily reacts with compounds containing amino groups, forming a chemically stable amide bond between the dye, e.g., a protein. The optimum pH-range for NHS-ester coupling is pH 8.0 – 9.0. At this pH amino groups of proteins, i.e., the ε-amino groups of lysines are unprotonated to a degree sufficiently high for fast coupling.

Features and Benefits

  • Strong absorption
  • High fluorescence quantum yield
  • High thermal and photo-stability

Other Notes

Fluorescence of bioconjugates; employed in fluorescence resonance energy transfer, FRET, experiments.

Legal Information

This product is for Research use only. In case of intended commercialization, please contact the IP-holder (ATTO-TEC GmbH, Germany) for licensing.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Benjamin Zoller et al.
Cell, 175(3), 835-847 (2018-10-20)
How transcriptional bursting relates to gene regulation is a central question that has persisted for more than a decade. Here, we measure nascent transcriptional activity in early Drosophila embryos and characterize the variability in absolute activity levels across expression boundaries.
Michael Cooper et al.
Journal of fluorescence, 14(2), 145-150 (2004-12-24)
The spectral properties of a rigidified trimethine cyanine dye, Cy3B have been characterised. This probe has excellent fluorescent properties, good water solubility and can be bioconjugated. The emission properties of this fluorophore have also been investigated upon conjugation to an
Elizabeth A Jares-Erijman et al.
Nature biotechnology, 21(11), 1387-1395 (2003-11-05)
Förster (or Fluorescence) Resonance Energy Transfer (FRET) is unique in generating fluorescence signals sensitive to molecular conformation, association, and separation in the 1-10 nm range. We introduce a revised photophysical framework for the phenomenon and provide a systematic catalog of
Rachael Bakker et al.
eLife, 9 (2020-06-23)
Morphogen signaling contributes to the patterned spatiotemporal expression of genes during development. One mode of regulation of signaling-responsive genes is at the level of transcription. Single-cell quantitative studies of transcription have revealed that transcription occurs intermittently, in bursts. Although the
Matthew G Niepielko et al.
Current biology : CB, 28(12), 1872-1881 (2018-06-05)
The formation of ribonucleoprotein assemblies called germ granules is a conserved feature of germline development. In Drosophila, germ granules form at the posterior of the oocyte in a specialized cytoplasm called the germ plasm, which specifies germline fate during embryogenesis. mRNAs

Articles

Chromogenic and fluorogenic derivatives are invaluable tools for biochemistry, having numerous applications in enzymology, protein chemistry, immunology and histochemistry.

Chromogenic and fluorogenic derivatives are invaluable tools for biochemistry, having numerous applications in enzymology, protein chemistry, immunology and histochemistry.

Chromogenic and fluorogenic derivatives are invaluable tools for biochemistry, having numerous applications in enzymology, protein chemistry, immunology and histochemistry.

Chromogenic and fluorogenic derivatives are invaluable tools for biochemistry, having numerous applications in enzymology, protein chemistry, immunology and histochemistry.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service