Skip to Content
Merck
All Photos(1)

Key Documents

69042

Sigma-Aldrich

Strontium chloride solution

0.1 M

Synonym(s):

Additive Screening Solution 27/Kit-No 78374

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.32

form

liquid

Quality Level

concentration

0.1 M

density

1.01 g/mL at 20 °C

storage temp.

2-8°C

SMILES string

[Sr]

InChI

1S/Sr

InChI key

CIOAGBVUUVVLOB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Strontium chloride solution is an additive screening solution of Additive Screening Kit. Additive Screen kit is designed to allow rapid and convenient evaluation of additives and their ability to influence the crystallization of the sample. The Additive Kit provides a tool for refining crystallization conditions.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gang Liu et al.
Theriogenology, 113, 78-84 (2018-02-24)
The survival rate of vitrified-thawed ovarian tissues after autotransplantation still needs to be improved. Therefore finding an ideal cryoprectant to reduce the damage to ovaries that caused by vitrification will pave the way for application of ovary cryopreservation on clinics.
Serdal Kirmizialtin et al.
Biophysical journal, 102(4), 829-838 (2012-03-06)
The distributions of different cations around A-RNA are computed by Poisson-Boltzmann (PB) equation and replica exchange molecular dynamics (MD). Both the nonlinear PB and size-modified PB theories are considered. The number of ions bound to A-RNA, which can be measured
D K Fomin et al.
Vestnik rentgenologii i radiologii, (2)(2), 29-31 (2012-06-27)
To study the efficiency of treatment via single administration of high-purity 89Sr chloride in the standard activity of 150 MBq for pain syndrome in patients with multiple bone metastases. The authors carried out clinical trials of high-purity 89Sr chloride used
Karen A Autio et al.
Current treatment options in oncology, 13(2), 174-188 (2012-04-25)
Skeletal metastases threaten quality of life, functionality, and longevity in patients with metastatic castration-resistant prostate cancer (mCRPC). Therapeutic strategies for bone metastases in prostate cancer can palliate pain, delay/prevent skeletal complications, and prolong survival. Pharmacologic agents representing several drug classes
Diane Cummins
American journal of dentistry, 23 Spec No A, 3A-13A (2011-02-03)
To provide a brief overview of the diagnosis, epidemiology, etiology and clinical management of dentin hypersensitivity, to discuss technical approaches to relieve sensitivity, with special emphasis on dentin tubule occlusion and the clinical evidence for efficacy of desensitizing toothpastes based

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service