Skip to Content
Merck
All Photos(1)

Key Documents

59331C

SAFC

Dulbecco′s Phosphate Buffered Saline

liquid, sterile-filtered, DPBS Modified 10X, without calcium, without magnesium, suitable for cell culture

Pharma Manufacturing

Synonym(s):

DPBS

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352207
NACRES:
NA.75
Pricing and availability is not currently available.

Quality Level

description

for research or for further manufacturing use

sterility

sterile-filtered

form

liquid

technique(s)

cell culture | mammalian: suitable

shipped in

ambient

Looking for similar products? Visit Product Comparison Guide

Application

Dulbecco′s Phosphate Buffered Saline (DPBS) is intended for use in the maintenance of mammalian cells where a chemically defined balanced salt solution provides an environment that will maintain the structural and physiological integrity of cells in vitro. DPBS is commonly used in cell enumerations as a diluent, for rinsing cells and as a buffer in many chromatographic procedures. DPBS also is used to wash and resuspend cells during the dissociation process, where the presence of calcium and magnesium can inhibit trypsin activity. For this application, use DPBS Modified, which is formulated without calcium and magnesium salts.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Amir Tirosh et al.
Science translational medicine, 11(489) (2019-04-26)
The short-chain fatty acid propionate is a potent inhibitor of molds that is widely used as a food preservative and endogenously produced by gut microbiota. Although generally recognized as safe by the U.S. Food and Drug Administration, the metabolic effects

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service