Skip to Content
Merck
All Photos(1)

Key Documents

56968

Sigma-Aldrich

Sparfloxacin

≥98.0% (HPLC)

Synonym(s):

5-Amino-1-cyclohexyl-7-(cis-3,5-dimethylpiperazino)-6,8-difluoro-1,4-dihydro-4-oxo-3-quinolinecarboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H22F2N4O3
CAS Number:
Molecular Weight:
392.40
Beilstein:
9170271
MDL number:
UNSPSC Code:
51282944
PubChem Substance ID:
NACRES:
NA.85

biological source

synthetic

Assay

≥98.0% (HPLC)

form

powder

color

white to light yellow

antibiotic activity spectrum

Gram-negative bacteria
Gram-positive bacteria
mycobacteria
mycoplasma

Mode of action

enzyme | inhibits

SMILES string

C[C@H]1CN(C[C@@H](C)N1)c2c(F)c(N)c3C(=O)C(=CN(C4CC4)c3c2F)C(O)=O

InChI

1S/C19H22F2N4O3/c1-8-5-24(6-9(2)23-8)17-13(20)15(22)12-16(14(17)21)25(10-3-4-10)7-11(18(12)26)19(27)28/h7-10,23H,3-6,22H2,1-2H3,(H,27,28)/t8-,9+

InChI key

DZZWHBIBMUVIIW-DTORHVGOSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Application

Sparfoxacin is used to study antimicrobial activity against Mycobacteria and respiratory tract infections .

Biochem/physiol Actions

Sparfloxacin fluoroquinolone antibiotic that inhibits bacterial DNA gyrase (topoisomerase II) and/or topoisomerase IV. It inhibits bacterial DNA gyrase-dependent processes such as DNA polymerization, ATP-dependent-DNA supercoiling, and chromosome fragmentation.

Packaging

1g, 10g

Other Notes

Keep container tightly closed in a dry and well-ventilated place. Keep in a dry place.Storage class (TRGS 510): Non Combustible Solids

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

N Rastogi et al.
Antimicrobial agents and chemotherapy, 35(12), 2473-2480 (1991-12-01)
The MICs and MBCs of the new difluorinated quinolone drug sparfloxacin against type strains belonging to 21 species of mycobacteria were screened. The MICs and MBCs were within the range of 0.1 to 2.0 and 0.1 to 4.0 micrograms/ml, respectively
K L Goa et al.
Drugs, 53(4), 700-725 (1997-04-01)
Sparfloxacin is a fluoroquinolone antibacterial agent with activity against a broad range of Gram-negative and Gram-positive organisms including Streptococcus pneumoniae, one of the main pathogens in community-acquired pneumonia. In this infection, sparfloxacin has shown efficacy similar to that of amoxicillin
Kenichiro Shimizu et al.
Antimicrobial agents and chemotherapy, 52(10), 3823-3825 (2008-07-23)
A new chromosome-carried quinolone resistance gene from Stenotrophomonas maltophilia, Smqnr, was characterized. The gene was present in type strain CCUG 5866 and was also detected in 24 clinical isolates and showed some allelic diversity. The expression of Smqnr in Escherichia
Sonia K Morgan-Linnell et al.
Antimicrobial agents and chemotherapy, 51(11), 4205-4208 (2007-08-08)
In defined, isogenic strains, at least three mutations, two of which must be in gyrA, were required to exceed the CLSI breakpoint for fluoroquinolone resistance. Strains with double mutations in both gyrA and parC had even higher MICs of fluoroquinolones
Radha K Shandil et al.
Antimicrobial agents and chemotherapy, 51(2), 576-582 (2006-12-06)
Members of the fluoroquinolone class are being actively evaluated for inclusion in tuberculosis chemotherapy regimens, and we sought to determine the best in vitro and pharmacodynamic predictors of in vivo efficacy in mice. MICs for Mycobacterium tuberculosis H37Rv were 0.1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service