Skip to Content
Merck
All Photos(1)

Documents

52976

Sigma-Aldrich

Kuromanin chloride

≥95% (HPLC)

Synonym(s):

Asterin, Chrysanthemin, Cyanidin 3-O-glucoside chloride, Glucocyanidin chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H21ClO11
CAS Number:
Molecular Weight:
484.84
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.25

Quality Level

Assay

≥95% (HPLC)

storage temp.

−20°C

SMILES string

O[C@@H]1[C@@H](O)[C@H](OC2=CC3=C([O+]=C2C4=CC(O)=C(O)C=C4)C=C(O)C=C3O)O[C@H](CO)[C@H]1O.[Cl-]

InChI

1S/C21H20O11.ClH/c22-7-16-17(27)18(28)19(29)21(32-16)31-15-6-10-12(25)4-9(23)5-14(10)30-20(15)8-1-2-11(24)13(26)3-8;/h1-6,16-19,21-22,27-29H,7H2,(H3-,23,24,25,26);1H/t16-,17-,18+,19-,21-;/m1./s1

InChI key

YTMNONATNXDQJF-UBNZBFALSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Anthocyanin. Antioxidant. Studied for its neuroprotective property.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ne Long et al.
Cancer science, 104(3), 298-303 (2012-12-04)
Purple corn color is a widely used food colorant that was reported to have attenuating effects on hypertension, diabetes, and to have anti-cancer effects on colon and breast cancer. Our study is the first on its possible chemoprevention effects against
Dongliang Wang et al.
Molecular nutrition & food research, 56(4), 610-621 (2012-04-13)
Although previous studies have shown that consumption of anthocyanin extract from plant foods reduces hypercholesterolemia and the severity of atherosclerosis in different animal models, the mechanisms of these actions remained unclear. This study investigated whether pure anthocyanin inhibit atherosclerosis development
S Anwar et al.
Toxicology letters, 226(2), 206-213 (2014-02-13)
The term hypoxia refers to conditions characterized by a relative restriction of oxygen supply. It is usually associated to a paradoxical overproduction of reactive oxygen species (ROS) and to the activation of several transcription factors, including HIF-1α, which in turn
Zhou Zhou et al.
Phytomedicine : international journal of phytotherapy and phytopharmacology, 19(10), 878-881 (2012-06-19)
Studies have shown positive correlations between inflammatory cytokines such as interleukin-6 (IL-6) and the development of chronic diseases including cardiovascular disease by activating C-reactive protein (CRP). Both atorvastatin calcium (lipitor) as well as flavonoid rich fruit such as tart cherry
Jacynthe Desjardins et al.
Journal of medicinal food, 15(12), 1045-1050 (2012-06-29)
Periodontal diseases are a group of multifactorial polymicrobial infections characterized by a progressive inflammatory destruction of the periodontium. Flavonoids, including anthocyanins, are receiving increasing attention because of their promising human health benefits. The aim of our study was to investigate

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service