51845
1,2,3-Heptanetriol
high melting isomer, ≥98.0% (GC)
Synonym(s):
1,2,3-Trihydroxyheptane, Heptane-1,2,3-triol
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
CH3(CH2)3CH(OH)CH(OH)CH2OH
CAS Number:
Molecular Weight:
148.20
Beilstein:
1737083
MDL number:
UNSPSC Code:
12162007
PubChem Substance ID:
NACRES:
NA.26
Recommended Products
Quality Level
Assay
≥98.0% (GC)
form
solid
mp
52-55 °C (lit.)
79-81 °C
SMILES string
CCCCC(O)C(O)CO
InChI
1S/C7H16O3/c1-2-3-4-6(9)7(10)5-8/h6-10H,2-5H2,1H3
InChI key
HXYCHJFUBNTKQR-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Other Notes
Amphiphile used for the crystallization of membrane proteins. It is added prior to crystallization to the protein solubilized by a small detergent (LDAO or octyl glucoside); Crystallization of reaction centers
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
W. Welte et al.
Febs Letters, 182, 260-260 (1985)
Three-dimensional crystals of a membrane protein complex. The photosynthetic reaction centre from Rhodopseudomonas viridis.
H Michel
Journal of molecular biology, 158(3), 567-572 (1982-07-05)
H. Michel
Trends in Biochemical Sciences, 8, 56-56 (1983)
Rosaria Gandini et al.
Journal of molecular biology, 432(16), 4658-4672 (2020-06-23)
Protein glycosylation constitutes a critical post-translational modification that supports a vast number of biological functions in living organisms across all domains of life. A seemingly boundless number of enzymes, glycosyltransferases, are involved in the biosynthesis of these protein-linked glycans. Few
J P Allen et al.
Proceedings of the National Academy of Sciences of the United States of America, 81(15), 4795-4799 (1984-08-01)
Reaction centers (RCs), integral membrane proteins that mediate the conversion of light into chemical energy, were crystallized by two different vapor diffusion techniques. In one method, small amphipathic molecules (1,2,3-heptanetriol and triethylammonium phosphate) were added to the RCs that had
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service