Skip to Content
Merck
All Photos(3)

Key Documents

478M-9

Sigma-Aldrich

CDK4 (DCS-31) Mouse Monoclonal Antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

mouse

Quality Level

100
500

conjugate

unconjugated

antibody form

culture supernatant

antibody product type

primary antibodies

clone

DCS-31, monoclonal

description

For In Vitro Diagnostic Use in Select Regions

form

buffered aqueous solution

species reactivity

human

packaging

vial of 0.1 mL concentrate (478M-94)
vial of 0.1 mL concentrate Research Use Only (478M-94-RUO)
vial of 0.5 mL concentrate (478M-95)
vial of 1.0 mL concentrate (478M-96)
vial of 1.0 mL concentrate Research Use Only (478M-96-RUO)
vial of 1.0 mL pre-dilute Research Use Only (478M-97-RUO)
vial of 1.0 mL pre-dilute ready-to-use (478M-97)
vial of 7.0 mL pre-dilute ready-to-use (478M-98)
vial of 7.0 mL pre-dilute ready-to-use Research Use Only (478M-98-RUO)

manufacturer/tradename

Cell Marque

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:25-1:50 (concentrated)

isotype

IgG

control

dedifferentiated liposarcoma, well-differentiated liposarcoma

shipped in

wet ice

storage temp.

2-8°C

visualization

nuclear

General description

Well-differentiated liposarcoma is a malignant, non-metastasizing tumor that can present with similar histological characteristics as benign lipoma lesions and has a higher recurrence rate. The CDK4 protein is frequently overexpressed in well-differentiated liposarcoma due to gene amplification of the 12q13-15 chromosomal region that harbors the CDK4 gene, but amplification and subsequent overexpression is rarely observed in lipomas. Due to CDK4 protein expression differences between these lesions, anti-CDK4 antibody can be used in an immunohistochemistry panel as an aid in the differential diagnosis between well-differentiated liposarcoma and lipoma.1,2,3

Quality

United States - IVD
Canada - RUO
European Union - IVD
Japan - RUO

Physical form

Solution in Tris Buffer, pH 7.3-7.7, with 1% BSA and <0.1% Sodium Azide.

Preparation Note

Download the IFU specific to your product lot and format
Note: This requires a keycode which can be found on your packaging or product label.

Download the latest released IFU
Note: This IFU may not apply to your specific product lot.

Other Notes

For Technical Service please contact: 800-665-7284 or email: service@cellmarque.com

Legal Information

Cell Marque is a trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Matthieu Bui Nguyen Binh et al.
The American journal of surgical pathology, 29(10), 1340-1347 (2005-09-15)
Atypical lipomatous tumor/well-differentiated liposarcoma (ALT-WDLPS) and dedifferentiated liposarcoma (DDLPS) may be difficult to distinguish from benign adipose tumors and from poorly differentiated sarcomas, respectively. Genetically, they are characterized by amplification of MDM2 and CDK4 genes on chromosome 12q13-15. We examined
P B Aleixo et al.
Journal of clinical pathology, 62(12), 1127-1135 (2009-12-01)
Well differentiated liposarcomas (WDLPS) and dedifferentiated liposarcomas (DDLPS) have been shown to have supernumerary chromosomes with amplified sequences of the MDM2 and CDK4 genes. MDM2 and CDK4 protein overexpression have also been identified in these tumours. To investigate whether immunohistochemistry
Khin Thway et al.
The American journal of surgical pathology, 36(3), 462-469 (2012-02-04)
Adipocytic tumors are the most common type of soft tissue neoplasms. Distinguishing atypical lipomatous tumor-well-differentiated liposarcoma (WDL) from benign adipocytic neoplasms and dedifferentiated liposarcoma (DDL) from pleomorphic or myxoid liposarcoma (LPS) can be difficult. WDL and DDL characteristically harbor amplifications

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service