46393
Fluo-3
suitable for fluorescence, ~70%
Synonym(s):
4-(2,7-Dichloro-6-hydroxy-3-oxo-9-xanthenyl)-4′-methyl-2,2′-(ethylenedioxy)dianiline-N,N,N′,N′-tetraacetic acid
About This Item
Assay
~70%
Quality Level
form
solid
solubility
H2O: soluble
fluorescence
λex 506 nm; λem 526 nm in 0.1 M Tris pH 8.0, esterase 30 mM Ca2+
suitability
suitable for fluorescence
storage temp.
−20°C
SMILES string
Cc1ccc(N(CC(O)=O)CC(O)=O)c(OCCOc2cc(ccc2N(CC(O)=O)CC(O)=O)C3=C4C=C(Cl)C(=O)C=C4Oc5cc(O)c(Cl)cc35)c1
InChI
1S/C36H30Cl2N2O13/c1-18-2-4-24(39(14-32(43)44)15-33(45)46)30(8-18)51-6-7-52-31-9-19(3-5-25(31)40(16-34(47)48)17-35(49)50)36-20-10-22(37)26(41)12-28(20)53-29-13-27(42)23(38)11-21(29)36/h2-5,8-13,41H,6-7,14-17H2,1H3,(H,43,44)(H,45,46)(H,47,48)(H,49,50)
InChI key
OZLGRUXZXMRXGP-UHFFFAOYSA-N
Application
Other Notes
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service