Skip to Content
Merck
All Photos(1)

Documents

45763

Supelco

Fenazox

PESTANAL®, analytical standard

Synonym(s):

Azoxybenzene, Fenazox

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H10N2O
CAS Number:
Molecular Weight:
198.22
Beilstein:
7462853
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

application(s)

agriculture
environmental

format

neat

SMILES string

[O-]\[N+](=N/c1ccccc1)c2ccccc2

InChI

1S/C12H10N2O/c15-14(12-9-5-2-6-10-12)13-11-7-3-1-4-8-11/h1-10H/b14-13-

InChI key

GAUZCKBSTZFWCT-YPKPFQOOSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Kujawa et al.
Zeitschrift fur die gesamte Hygiene und ihre Grenzgebiete, 35(5), 255-257 (1989-05-01)
No sex differences could been shown after absorption and distribution of azoxybenzen in the rat. The highest level occur in the thyroid gland, the adrenals and in the fat tissues. The maximum level was reached 30 min. after application.
J P Tindall
Journal of the American Academy of Dermatology, 13(4), 539-558 (1985-10-01)
Chloracne, an acneform eruption resulting from poisoning by halogenated aromatic compounds, has been a considerable problem over the last 40 years. The condition is always a symptom of systemic poisoning and should be familiar to all practitioners, particularly dermatologists. It
R Plass et al.
Die Nahrung, 32(10), 989-997 (1988-01-01)
Oral administrations of azoxybenzene (0, 25, 50 and 100 mg/kg b.w.) over a period up to 7 days caused a time and dose dependent decrease in the amount of cytochrome P-450 as well as in the ability of aminopyrine-N-demethylation and
M Stock et al.
Die Nahrung, 29(5), 473-479 (1985-01-01)
Results from investigations on the metabolism of 14C-Fenazox show that in 6-8 weeks old tomato-plants (sort "Harzfeuer") the agent undergoes a biotransformation. After chromatographic separation the structure of the biotransformation products was elucidated by comparison to authentic test substances, by
Investigations of myelotoxic effects in rats.
T A Moeller
Archives of toxicology. Supplement. = Archiv fur Toxikologie. Supplement, 14, 83-87 (1991-01-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service