Skip to Content
Merck
All Photos(1)

Documents

45690

Sigma-Aldrich

Erythrosin extra bluish

for microscopy (Bact., Hist.), adsorption and fluorescent indicator

Synonym(s):

2′,4′,5′,7′-Tetraiodofluorescein disodium salt, Acid Red 51, Iodoeosin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H6I4Na2O5
CAS Number:
Molecular Weight:
879.86
Colour Index Number:
45430
Beilstein:
1443945
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

grade

adsorption and fluorescent indicator
for microscopy (Bact., Hist.)

Quality Level

form

solid

technique(s)

titration: suitable

SMILES string

[Na+].[Na+].[O-]c1c(I)cc2c(Oc3c(I)c([O-])c(I)cc3C24OC(=O)c5ccccc45)c1I

InChI

1S/C20H8I4O5.2Na/c21-11-5-9-17(13(23)15(11)25)28-18-10(6-12(22)16(26)14(18)24)20(9)8-4-2-1-3-7(8)19(27)29-20;;/h1-6,25-26H;;/q;2*+1/p-2

InChI key

RAGZEDHHTPQLAI-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Erythrosin extra bluish is a brown color powder, that is soluble in ethanol and water. Its pH ranges from 2.5 to 4. It is used in color filters, light emitting diodes, cosmetics, hair dyes etc. It has several biological applications like detecting gene expression, treating age related macular degeneration, diabetes, obesity etc.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

359.6 °F - closed cup

Flash Point(C)

182 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Juliana P M L Rolim et al.
Journal of photochemistry and photobiology. B, Biology, 106, 40-46 (2011-11-11)
Several photosensitizers have been used against oral bacteria without standardization. Singlet oxygen ((1)O(2)) is an aggressive chemical species that can kill cells through apoptosis or necrosis. to compare the antimicrobial activity of photodynamic therapy (PDT) with different photosensitizers at the
Jun Liang et al.
Carbohydrate research, 361, 120-126 (2012-09-29)
The effect of glycerol on molecular mobility and hydrogen bonding in amorphous glucose matrix was studied. Phosphorescence from erythrosin B (Ery B) was used to characterize the temperature dependence of mobility in glucose/glycerol films over the temperature range from 100
Anna Carolina Borges Pereira Costa et al.
Mycoses, 55(1), 56-63 (2011-06-15)
The aim of this study was to evaluate the effects of photodynamic therapy (PDT) using rose bengal or erythrosine with light emitting diode (LED) on Candida albicans planktonic cultures and biofilms. Seven C. albicans clinical strains and one standard strain
Satoshi Horikoshi et al.
Clinica chimica acta; international journal of clinical chemistry, 413(13-14), 1087-1091 (2012-04-03)
In spite of the urgent necessity for a screening test of urinary protein for the early diagnosis of kidney diseases, a rapid, accurate and cost-effective method for their detection has yet to be developed. A solution containing a buffer agent
Anna Carolina Borges Pereira Costa et al.
Oral surgery, oral medicine, oral pathology and oral radiology, 114(1), 67-74 (2012-06-26)
This study evaluated the effects of photodynamic therapy (PDT) on buccal candidiasis in mice and on the adherence of yeast to buccal epithelial cells (BECs) in vitro. A total of 56 immunosuppressed mice with buccal candidiasis were subjected to PDT

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service