Skip to Content
Merck
All Photos(2)

Key Documents

34340

Supelco

Etridiazole

PESTANAL®, analytical standard

Synonym(s):

5-Ethoxy-3-trichloromethyl-1,2,4-thiadiazole, Ethazol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H5Cl3N2OS
CAS Number:
Molecular Weight:
247.53
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

application(s)

agriculture
cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

SMILES string

CCOc1nc(ns1)C(Cl)(Cl)Cl

InChI

1S/C5H5Cl3N2OS/c1-2-11-4-9-3(10-12-4)5(6,7)8/h2H2,1H3

InChI key

KQTVWCSONPJJPE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Etridiazole is a fungicide, which is found to be effective against the oomycete group of fungi. It is used for seed treatment of various crops, in cotton cultivation during planting stage and non-bearing coffee and citrus fruits by horticultural nurseries.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

310.1 °F - open cup

Flash Point(C)

154.5 °C - open cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

[Severe manifestations of drug allergy in children].
T N Sukovatykh et al.
Pediatriia, (5)(5), 76-77 (1982-05-01)
Sittig's Handbook of Pesticides and Agricultural Chemicals (2013)
P P Golikov et al.
Voprosy meditsinskoi khimii, 40(2), 9-12 (1994-03-01)
The effect of sodium sulfaethidole on Types II and III glucocorticoid receptors was studied in liver cytosol fraction obtained from female Wistar rats of 180-200 g body weight. Functions of Type II glucocorticoid receptors in liver cytosol were evaluated by
A toxicologic evaluation of 5-ethoxy-3-trichloromethyl-1,2,4-thiadiazole (ETMT).
J F Borzelleca et al.
Toxicology and applied pharmacology, 56(2), 164-170 (1980-11-01)
[Effect of sulfonamides on the fatty acid composition of enterobacterial lipids].
Z P Vasiurenko et al.
Mikrobiologicheskii zhurnal, 47(1), 29-36 (1985-01-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service