Skip to Content
Merck
All Photos(1)

Key Documents

326M-1

Sigma-Aldrich

PSAP (PASE/4LJ) Mouse Monoclonal Antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

100
500

conjugate

unconjugated

antibody form

culture supernatant

antibody product type

primary antibodies

clone

PASE/4LJ, monoclonal

description

For In Vitro Diagnostic Use in Select Regions (See Chart)

form

buffered aqueous solution

species reactivity

human

packaging

vial of 0.1 mL concentrate (326M-14)
vial of 0.5 mL concentrate (326M-15)
bottle of 1.0 mL predilute (326M-17)
vial of 1.0 mL concentrate (326M-16)
bottle of 7.0 mL predilute (326M-18)

manufacturer/tradename

Cell Marque

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:25-1:100

isotype

IgG1

control

prostate

shipped in

wet ice

storage temp.

2-8°C

visualization

cytoplasmic

Gene Information

human ... PSAP(5660)

General description

Anti-PSAP reacts with prostatic acid phosphatase in the glandular epithelium of normal and hyperplastic prostate, carcinoma of the prostate, and metastatic cells of prostatic carcinoma. This marker may be helpful in pinpointing the site of origin in cases of metastatic carcinoma of the prostate, and is considered a more sensitive marker than PSA. However, it also offers less specificity. Nevertheless, PSAP complements PSA in the right clinical context.

Quality


IVD

IVD

IVD

RUO

Linkage

PSAP Positive Control Slides, Product No. 326S, are available for immunohistochemistry (formalin-fixed, paraffin-embedded sections).

Physical form

Solution in Tris Buffer, pH 7.3-7.7, with 1% BSA and <0.1% Sodium Azide

Preparation Note

Download the IFU specific to your product lot and formatNote: This requires a keycode which can be found on your packaging or product label.

Other Notes

For Technical Service please contact: 800-665-7284 or email: service@cellmarque.com

Legal Information

Cell Marque is a trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

N Kuroda et al.
Pathology international, 49(5), 457-461 (1999-07-27)
Signet-ring cell carcinoma (SRCC) of the prostate is a very rare neoplasm and there have been only 38 cases reported to date. Here the 39th case of prostatic SRCC containing a small amount of neutral mucin, prostatic specific antigen (PSA)
Noman Kidwai et al.
Breast cancer research : BCR, 6(1), R18-R23 (2003-12-19)
The androgen-regulated proteins prostate-specific antigen (PSA) and prostate-specific acid phosphatase (PSAP) are present in high concentrations in normal prostate and prostatic cancer and are considered to be tissue-specific to prostate. These markers are commonly used to diagnose metastatic prostate carcinoma
E M Genega et al.
Modern pathology : an official journal of the United States and Canadian Academy of Pathology, Inc, 13(11), 1186-1191 (2000-12-06)
Morphologic features alone can usually be used to distinguish prostatic adenocarcinoma and urothelial carcinoma of the urinary bladder. Poorly differentiated tumors, however, can occasionally have features of both neoplasms, making determination of site of origin difficult. No study has provided
J I Epstein
The Urologic clinics of North America, 20(4), 757-770 (1993-11-01)
This article describes the immunoreactivity of PSA and PAP in non-neoplastic and neoplastic prostate tissue. Listed are examples of cross reactivity of PSA and PAP in non-neoplastic and neoplastic tissue from other organs. The use of PSA and PAP to
M A Ansari et al.
American journal of clinical pathology, 76(1), 94-98 (1981-07-01)
An unusual case of carcinoma of the prostate with metastases is described. the prostate and the metastases showed adenocarcinoma with carcinoid-like areas. A tumor with the same histologic features was found at the tip of the appendix and proved to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service