Skip to Content
Merck
All Photos(1)

Key Documents

318A-1

Sigma-Aldrich

PGP 9.5 Rabbit Polyclonal Antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

100
500

conjugate

unconjugated

antibody form

Ig fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

description

For In Vitro Diagnostic Use in Select Regions (See Chart)

form

buffered aqueous solution

species reactivity

human

packaging

vial of 0.1 mL concentrate (318A-14)
vial of 0.5 mL concentrate (318A-15)
bottle of 1.0 mL predilute (318A-17)
vial of 1.0 mL concentrate (318A-16)
bottle of 7.0 mL predilute (318A-18)

manufacturer/tradename

Cell Marque®

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:100-1:500

control

nerve tissue

shipped in

wet ice

storage temp.

2-8°C

visualization

cytoplasmic

General description

Protein gene product 9.5 (PGP 9.5), also known as ubiquitin carboxyl-terminal hydrolase-1 (UCHL-1), is a 27 kDa protein originally isolated from whole brain extracts. Although PGP 9.5 expression in normal tissues was originally felt to be strictly confined to neurons and neuroendocrine cells, it has been subsequently documented in distal renal tubular epithelium, spermatogonia, Leydig cells, oocytes, melanocytes, prostatic secretory epithelium, ejaculatory duct cells, epididymis, mammary epithelial cells, Merkel cells, and dermal fibroblasts. A plethora of mesenchymal neoplasms have demonstrated PGP 9.5 expression.

Quality


IVD

IVD

IVD

RUO

Linkage

PGP9.5 Positive Control Slides, Product No. 318S, are available for immunohistochemistry (formalin-fixed, paraffin-embedded sections).

Physical form

Solution in Tris Buffer, pH 7.3-7.7, with 1% BSA and <0.1% Sodium Azide

Preparation Note

Download the IFU specific to your product lot and formatNote: This requires a keycode which can be found on your packaging or product label.

Other Notes

For Technical Service please contact: 800-665-7284 or email: [email protected]

Legal Information

Cell Marque is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lucas K Campbell et al.
Modern pathology : an official journal of the United States and Canadian Academy of Pathology, Inc, 16(10), 963-969 (2003-10-16)
In non-neoplastic tissues, the expression of protein gene product 9.5 (PGP 9.5), a member of the ubiquitin hydrolase family of proteins, is confined to neural and neuroendocrine cells. Although it has been claimed that PGP 9.5 is a specific marker
G Bassotti et al.
Journal of clinical pathology, 58(9), 973-977 (2005-08-30)
Colonic diverticular disease (diverticulosis) is a common disorder in Western countries. Although its pathogenesis is probably multifactorial, motor abnormalities of the large bowel are thought to play an important role. However, little is known about the basic mechanism that may
M Mahalingam et al.
Journal of cutaneous pathology, 33(1), 51-56 (2006-01-31)
Cellular neurothekeoma is a relatively rare, benign cutaneous neoplasm, which usually presents as a solitary papule or nodule involving the head and neck area of young adults. Multiple neurothekeomas have not, to date, been known to occur. We report a
M Mahalingam et al.
Journal of cutaneous pathology, 28(6), 282-286 (2001-06-13)
Protein gene product 9.5 (PGP 9.5) is expressed in brain at 20 to 50 times the levels detected in other organs. Immunohistochemical studies reveal this protein is localized to both central and peripheral neurons. Recently, PGP 9.5 is reported to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service