Skip to Content
Merck
All Photos(1)

Key Documents

30450

Sigma-Aldrich

Daunorubicin hydrochloride

≥90% (HPLC)

Synonym(s):

Daunomycin hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H29NO10 · HCl
CAS Number:
Molecular Weight:
563.98
Beilstein:
4229221
EC Number:
MDL number:
UNSPSC Code:
51102829
PubChem Substance ID:
NACRES:
NA.85

biological source

synthetic

Assay

≥90% (HPLC)

form

powder

color

red to deep red

antibiotic activity spectrum

neoplastics

Mode of action

DNA synthesis | interferes

storage temp.

2-8°C

SMILES string

Cl.COc1cccc2C(=O)c3c(O)c4C[C@](O)(C[C@H](O[C@H]5C[C@H](N)[C@H](O)[C@H](C)O5)c4c(O)c3C(=O)c12)C(C)=O

InChI

1S/C27H29NO10.ClH/c1-10-22(30)14(28)7-17(37-10)38-16-9-27(35,11(2)29)8-13-19(16)26(34)21-20(24(13)32)23(31)12-5-4-6-15(36-3)18(12)25(21)33;/h4-6,10,14,16-17,22,30,32,34-35H,7-9,28H2,1-3H3;1H/t10-,14-,16-,17-,22+,27-;/m0./s1

InChI key

GUGHGUXZJWAIAS-QQYBVWGSSA-N

Gene Information

human ... TOP2A(7153)

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: aminoglycoside

Application

Daunorubicin hydrochloride is used in photostability, antileukemic, and drug metabolism studies.

Biochem/physiol Actions

On binding to DNA, daunomycin intercalates, with its daunosamine residue directed toward the minor groove. Daunomycin effectively binds to every 3 base pairs which causes unwinding.
Potent anticancer agent. Inhibits DNA and RNA synthesis as sequence specific ds-DNA intercalating agent.

Packaging

5 mg, 25 mg

Other Notes

Keep container tightly closed in a dry and well-ventilated place. Hygroscopic. Store under inert gas.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Carc. 2 - Muta. 2 - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Photoprotection of Daunorubicin Hydrochloride with Sodium Sulfite.
Mohammad S. Islam* and Ahmed F. Asker
Journal of Pharmaceutical Sciences, 49, 122-126 (1995)
Jay Chhablani et al.
Investigative ophthalmology & visual science, 54(2), 1268-1279 (2013-01-17)
To test the feasibility of covalent loading of daunorubicin into oxidized porous silicon (OPS) and to evaluate the ocular properties of sustained delivery of daunorubicin in this system. Porous silicon was heat oxidized and chemically functionalized so that the functional
Sarah Bertoli et al.
Blood, 121(14), 2618-2626 (2013-02-01)
In acute myeloid leukemia (AML), new strategies assess the potential benefit of genetically targeted therapy at diagnosis. This implies waiting for laboratory tests and therefore a delay in initiation of chemotherapy. We studied the impact of time from diagnosis to
Kevin W H Yee et al.
Blood, 104(13), 4202-4209 (2004-08-12)
Fetal liver tyrosine kinase 3 internal tandem duplication (FLT3 ITD) mutations are the most common molecular abnormality associated with adult acute myeloid leukemia (AML). To exploit this molecular target, a number of potent and specific FLT3 kinase inhibitors have been
Oliver Teuffel et al.
British journal of haematology, 161(2), 192-203 (2013-02-13)
This systematic review and meta-analysis compared the efficacy of different anthracyclines and anthracycline dosing schedules for induction therapy in acute myeloid leukaemia in children and adults younger than 60 years of age. Twenty-nine randomized controlled trials were eligible for inclusion in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service