Skip to Content
Merck
All Photos(1)

Key Documents

30398

Sigma-Aldrich

Cytochrome c from bovine heart

≥95% (GE)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.61
Pricing and availability is not currently available.

biological source

bovine heart

Quality Level

Assay

≥95% (GE)

form

powder or crystals

mol wt

Mr ~13000

technique(s)

activity assay: suitable

impurities

≤5% cytochrome c reduced

solubility

H2O: 10 mg/mL, clear, dark red-brown

UniProt accession no.

storage temp.

−20°C

Gene Information

Looking for similar products? Visit Product Comparison Guide

Preparation Note

Prepared by a procedure using TCA.

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alexander N Volkov et al.
Biochemistry, 52(13), 2165-2175 (2013-03-23)
Here we present the preparation, biophysical characterization, and nuclear magnetic resonance (NMR) spectroscopy study of yeast cytochrome c peroxidase (CcP) constructs with enhanced solubility. Using a high-yield Escherichia coli expression system, we routinely produced uniformly labeled [(2)H,(13)C,(15)N]CcP samples with high
Margareta R A Blomberg et al.
Biochimica et biophysica acta, 1827(7), 826-833 (2013-04-27)
The membrane-bound enzyme cNOR (cytochrome c dependent nitric oxide reductase) catalyzes the reduction of NO in a non-electrogenic process. This is in contrast to the reduction of O2 in cytochrome c oxidase (CcO), the other member of the heme-copper oxidase
Shujun Yuan et al.
Biochemistry, 52(13), 2319-2327 (2013-03-26)
Apoptosome assembly is highly regulated in the intrinsic cell death pathway. To better understand this step, we created an improved model of the human apoptosome using a crystal structure of full length Apaf-1 and a single particle, electron density map
Gongquan Li et al.
Biochemical and biophysical research communications, 434(4), 809-814 (2013-04-25)
Small-molecule Bcl-2/Bcl-xL inhibitor Navitoclax represents a promising cancer therapeutic since preclinical and clinical studies with Navitoclax have demonstrated strong anticancer activity in several types of cancers. However, because Navitoclax has a low binding affinity to Mcl-1, anticancer activity by Navitoclax
M Harjai et al.
Anaesthesia and intensive care, 41(2), 175-183 (2013-03-28)
Sepsis remains as a leading cause of death in critically ill patients. Unfortunately, there have been very few successful specific therapeutic agents that can significantly reduce the attributable mortality and morbidity of sepsis. Developing novel therapeutic strategies to improve outcomes

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service