Skip to Content
Merck
All Photos(1)

Key Documents

30024

Sigma-Aldrich

Cyclosporin A

97.0-101.5% (on dried basis)

Synonym(s):

Antibiotic S 7481F1, Cyclosporine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C62H111N11O12
CAS Number:
Molecular Weight:
1202.61
Beilstein:
3647785
MDL number:
UNSPSC Code:
51102829
PubChem Substance ID:
NACRES:
NA.76
Pricing and availability is not currently available.

Quality Level

Assay

97.0-101.5% (on dried basis)

form

powder

color

white to off-white

antibiotic activity spectrum

fungi

Mode of action

enzyme | inhibits

storage temp.

2-8°C

SMILES string

CC[C@@H]1NC(=O)[C@H]([C@H](O)[C@H](C)C\C=C\C)N(C)C(=O)[C@H](C(C)C)N(C)C(=O)[C@H](CC(C)C)N(C)C(=O)[C@H](CC(C)C)N(C)C(=O)[C@@H](C)NC(=O)[C@H](C)NC(=O)[C@H](CC(C)C)N(C)C(=O)[C@@H](NC(=O)[C@H](CC(C)C)N(C)C(=O)CN(C)C1=O)C(C)C

InChI

1S/C62H111N11O12/c1-25-27-28-40(15)52(75)51-56(79)65-43(26-2)58(81)67(18)33-48(74)68(19)44(29-34(3)4)55(78)66-49(38(11)12)61(84)69(20)45(30-35(5)6)54(77)63-41(16)53(76)64-42(17)57(80)70(21)46(31-36(7)8)59(82)71(22)47(32-37(9)10)60(83)72(23)50(39(13)14)62(85)73(51)24/h25,27,34-47,49-52,75H,26,28-33H2,1-24H3,(H,63,77)(H,64,76)(H,65,79)(H,66,78)/b27-25+/t40-,41+,42-,43+,44+,45+,46+,47+,49+,50+,51+,52-/m1/s1

InChI key

PMATZTZNYRCHOR-CGLBZJNRSA-N

Gene Information

human ... PPIA(5478)

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: peptide

Application

Cyclosporin A is used for immunosuppressive studies [1][2].

Biochem/physiol Actions

A fungal metabolite possessing potent immunosuppressive properties. It inhibits the T-cell receptor signal transduction pathway via the formation of cyclosporin A−cyclophilin complex that inhibits calcineurin (protein phosphatase 2B). Inhibits nitric oxide synthesis induced by interleukin 1α, lipopolysaccharides and TNFα. Can block cytochrome c release from mitochondria.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

Keep container tightly closed in a dry and well-ventilated place. Keep in a dry place.Storage class (TRGS 510): Non-combustible, acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects.

related product

Product No.
Description
Pricing

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Carc. 1B - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

C Frieke Kuper et al.
Toxicologic pathology, 35(2), 226-232 (2007-03-17)
The effects of cyclosporin A (CY) and cyclophosphamide (CPS) on Peyer's patches (PP) were studied in Wistar rats, exposed in utero and neonatally or during adult age. In one study, pregnant dams received 5 or 15 mg/kg bw/day CY from
Valerie Waters et al.
American journal of respiratory cell and molecular biology, 33(2), 138-144 (2005-05-10)
Cyclosporin A (CsA) blocks T cell activation by interfering with the Ca2+-dependent phosphatase, calcineurin. Proinflammatory responses to bacteria that are activated by Ca2+-fluxes in airway cells are a potential target for CsA. Although local immunosuppression may be advantageous to control
Mi-Yeon Jang et al.
Journal of medicinal chemistry, 54(2), 655-668 (2010-12-22)
Herein we describe the synthesis and in vitro and in vivo activity of thiazolo[5,4-d]pyrimidines as a novel class of immunosuppressive agents, useful for preventing graft rejection after organ transplantation. This research resulted in the discovery of a series of compounds
Xiao-Ling Liu et al.
Bioorganic & medicinal chemistry, 16(1), 171-180 (2007-10-30)
A library of chalcones with basic functionalities were screened for inhibition of P-glycoprotein (Pgp, ABCB1) by the calcein-AM accumulation assay on MDCKII/MDR1 cells. Three members that had ring A substituted with 5-(1-ethylpiperidin-4-yl) and 2,4-dimethoxy groups were found to increase calcein-AM
Lotte Coelmont et al.
Antimicrobial agents and chemotherapy, 53(3), 967-976 (2008-12-24)
Debio 025 is a potent inhibitor of hepatitis C virus (HCV) replication (J. Paeshuyse et al., Hepatology 43:761-770, 2006). In phase I clinical studies, monotherapy (a Debio 025 dose of 1,200 mg twice a day) resulted in a mean maximal

Articles

Nitric oxide (NO) as a signal transporter in neurons, endothelial cells and in the immune system.

Questions

  1. What do you use to reconstitute cyclosporine? Ethanol or DMSO?

    1 answer
    1. This product can be reconstituted in the following:

      Dichloromethane: 10 mg/mL
      Ethanol: 10 mg/mL
      DMSO: 50 mg/mL
      Chloroform: 6 mg/mL
      H2O: insoluble

      Solutions may be stored at -20 °C protected from light.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service