Skip to Content
Merck
All Photos(1)

Key Documents

296R-1

Sigma-Aldrich

Myogenin (EP162) Rabbit Monoclonal Primary Antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

100
500

conjugate

unconjugated

antibody form

culture supernatant

antibody product type

primary antibodies

clone

EP162, monoclonal

description

(For In Vitro Diagnostic Use in Select Regions (See Chart))

form

buffered aqueous solution

species reactivity

human

packaging

bottle of 1.0 mL predilute (296R-17)
bottle of 7.0 mL predilute (296R-18)
vial of 0.1 mL concentrate (296R-14)
vial of 0.5 mL concentrate (296R-15)
vial of 1.0 mL concentrate (296R-16)
vial of 0.1 mL concentrate Research Use Only (296R-14-RUO)
vial of 1.0 mL concentrate Research Use Only (296R-16-RUO)
vial of 1.0 mL pre-dilute Research Use Only (296R-17-RUO)
vial of 7.0 mL pre-dilute ready-to-use Research Use Only (296R-18-RUO)

manufacturer/tradename

Cell Marque

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:25-1:100

isotype

IgG

control

rhabdomyosarcoma

shipped in

wet ice

storage temp.

2-8°C

visualization

nuclear

Gene Information

human ... MYOG(4656)

General description

Anti-myogenin labels the nuclei of myoblasts in developing muscle tissue, and is expressed in tumor cell nuclei of rhabdomyosarcoma and some leiomyosarcomas. Positive nuclear staining may occur in Wilms′ tumor.

Quality


IVD

IVD

IVD

RUO

Linkage

Myogenin Positive Control Slides, Product No. 296S, are available for immunohistochemistry (formalin-fixed, paraffin-embedded sections).

Physical form

Solution in Tris Buffer, pH 7.3-7.7, with 1% BSA and <0.1% Sodium Azide.

Preparation Note

Download the IFU specific to your product lot and formatNote: This requires a keycode which can be found on your packaging or product label.

Other Notes

For Technical Service please contact: 800-665-7284 or email: service@cellmarque.com

Legal Information

Cell Marque is a trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J B Miller
The Journal of cell biology, 111(3), 1149-1159 (1990-09-01)
Different mouse muscle cell lines were found to express distinct patterns of myosin heavy chain (MHC) isoforms, MyoD1, and myogenin, but there appeared to be no correlation between the pattern of MHC expression and the patterns of MyoD1 and myogenin
S Cui et al.
Pathology international, 49(1), 62-68 (1999-05-05)
New monoclonal anti-MyoD1 and anti-myogenin antibodies were evaluated immunohistochemically to determine whether they are useful in discriminating rhabdomyosarcoma (RMS) from other soft tissue tumors in routinely processed sections. Neither MyoD1 nor myogenin was expressed in normal, mature striated muscle. In
N P Wang et al.
The American journal of pathology, 147(6), 1799-1810 (1995-12-01)
The distinction of rhabdomyosarcoma (RMS) from other small blue round cell tumors of childhood, such as Ewing's sarcoma/peripheral primitive neuroectodermal tumor (pPNET) and neuroblastoma, continues to present a diagnostic challenge to pathologists. The recent recognition of the master role of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service