Skip to Content
Merck
All Photos(1)

Key Documents

27883

Sigma-Aldrich

Coumestrol

BioReagent, suitable for fluorescence, ≥97.5% (HPLC)

Synonym(s):

7,12-Dihydroxycoumestan

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H8O5
CAS Number:
Molecular Weight:
268.22
Beilstein:
266702
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.77

product line

BioReagent

Assay

≥97.5% (HPLC)

form

powder

mp

≥350 °C (lit.)

solubility

DMSO: soluble

fluorescence

λex 377 nm; λem 437 nm in 0.1 M Tris pH 8.0

suitability

suitable for fluorescence

SMILES string

Oc1ccc-2c(OC(=O)c3c-2oc4cc(O)ccc34)c1

InChI

1S/C15H8O5/c16-7-1-3-9-11(5-7)19-14-10-4-2-8(17)6-12(10)20-15(18)13(9)14/h1-6,16-17H

InChI key

ZZIALNLLNHEQPJ-UHFFFAOYSA-N

Gene Information

human ... ESR1(2099) , ESR2(2100)
mouse ... Esr1(13982)
rat ... Ar(24208)

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Coumestrol has anti-estrogenic actions in the brain and pituitary, mediated through estrogen receptor-α.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Man-Hai Liu et al.
Neurological research, 33(6), 663-672 (2011-06-29)
Estrogen replacement therapy can decrease the risk of developing Alzheimer's disease. Phytoestrogens have been proposed as potential alternatives to estrogen replacement therapy. The purpose of this study was to evaluate the in vitro protective effects of coumestrol on mice astrocytes.
Sarah J Lowry et al.
Nutrition and cancer, 64(6), 783-789 (2012-08-07)
Some forms of estrogen are associated with breast cancer risk as well as with mammographic density (MD), a strong marker of breast cancer risk. Whether phytoestrogen intake affects breast density, however, remains unclear. We evaluated the association between serum levels
Corinne C Hoerger et al.
Journal of agricultural and food chemistry, 59(3), 847-856 (2011-01-06)
We developed and validated three different sample preparation and extraction methods followed by HPLC-MS/MS (negative electrospray ionization) analysis for the quantification of estrogenic isoflavones (formononetin, daidzein, equol, biochanin A, and genistein) and coumestrol in red clover, soil, and manure. Plant
Elisa V Bandera et al.
BMC women's health, 11, 40-40 (2011-09-29)
While there is extensive literature evaluating the impact of phytoestrogen consumption on breast cancer risk, its role on ovarian cancer has received little attention. We conducted a population-based case-control study to evaluate phytoestrogen intake from foods and supplements and epithelial
D F Argenta et al.
Die Pharmazie, 66(12), 929-932 (2012-02-09)
A simple, rapid, and sensitive LC method to determine coumestrol incorporated in the lipid nanoemulsions was validated. The analyses were performed at room temperature on a reversed-phase C18 column using a mobile phase composed of methanol/water with 0.1% trifluoracetic acid

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service