Skip to Content
Merck
All Photos(1)

Documents

275R-1

Sigma-Aldrich

Ki-67 (SP6) Rabbit Monoclonal Antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

100
500

conjugate

unconjugated

antibody form

culture supernatant

antibody product type

primary antibodies

clone

SP6, monoclonal

description

For In Vitro Diagnostic Use in Select Regions (See Chart)

form

buffered aqueous solution

species reactivity

human

packaging

vial of 0.1 mL concentrate (275R-14)
vial of 0.5 mL concentrate (275R-15)
bottle of 1.0 mL predilute (275R-17)
vial of 1.0 mL concentrate (275R-16)
bottle of 7.0 mL predilute (275R-18)

manufacturer/tradename

Cell Marque

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:100-1:500

isotype

IgG

control

tonsil

shipped in

wet ice

storage temp.

2-8°C

visualization

nuclear

Gene Information

human ... MKI67(4288)

General description

The Ki-67 antigen is a nuclear, non-histone protein that is present in proliferatingcells. Though its specific role is largely postulated and still yet to be determined,Ki-67’s presence during cellular division makes it an excellent target for labeling rapidlygrowing malignancies. Given that its presence correlates with cell proliferation, theimmunohistochemical labeling of Ki-67 antigen may be useful in characterizing a lesion’sgrowth fraction.

Quality


IVD

IVD

IVD

RUO

Linkage

Ki-67 Positive Control Slides, Product No. 275S, are available for immunohistochemistry (formalin-fixed, paraffin-embedded sections).

Physical form

Solution in Tris Buffer, pH 7.3-7.7, with 1% BSA and <0.1% Sodium Azide

Preparation Note

Download the IFU specific to your product lot and formatNote: This requires a keycode which can be found on your packaging or product label.

Other Notes

For Technical Service please contact: 800-665-7284 or email: service@cellmarque.com

Legal Information

Cell Marque is a trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ran Marciano et al.
Cancers, 11(2) (2019-02-02)
Tumor cells utilize glucose to fuel their anabolic needs, including rapid proliferation. However, due to defective vasculature and increased glucose uptake, tumor cells must overcome glucose deprivation. Accordingly, tumor cells depend on cellular pathways promoting survival under such conditions. Targeting
Swerdlow S, et al. WHO clarifications of tumours of hematopoietic and lymphoid tissues. 4th edition, 2008. P 220.
WHO Classification of Tumours of Haematopoietic and Lymphoid Tissue, 220-220 (2008)
S W Coons et al.
Neurosurgery, 41(4), 878-884 (1997-10-08)
The prognostic significance of quantitative measurement of tumor proliferative activity was evaluated for oligodendroglial tumors. Ki-67/MIB-1 immunochemistry was used to measure proliferative activity in 81 oligodendrogliomas and oligoastrocytomas. The relationship among survival, proliferation, histological features, and clinical variables were evaluated
Nirmala Pathmanathan et al.
Journal of clinical pathology, 66(6), 512-516 (2013-02-26)
New approaches to the prognostic assessment of breast cancer have come from molecular profiling studies. A major feature of this work has been to emphasise the importance of cancer cell proliferation as a key discriminative indicator of recurrence risk for
Carmen J Allegra et al.
Journal of clinical oncology : official journal of the American Society of Clinical Oncology, 21(2), 241-250 (2003-01-15)
To define the value of thymidylate synthase (TS), Ki-67, and p53 as prognostic markers in patients with stage II and III colon carcinoma. We retrospectively analyzed the prognostic value of TS, Ki-67, and p53 in 706 patients with Dukes' B

Articles

Colorectal cancer is the third most common cancer in both men and women. An estimated 136,000 cases of colorectal cancer are expected to occur in 2016.

Protocols

3D cell culture protocol for generating epidermal human skin tissue using primary human keratinocytes, dermal fibroblasts, and collagen-coated transwell inserts.

3D cell culture protocol for generating epidermal human skin tissue using primary human keratinocytes, dermal fibroblasts, and collagen-coated transwell inserts.

3D cell culture protocol for generating epidermal human skin tissue using primary human keratinocytes, dermal fibroblasts, and collagen-coated transwell inserts.

3D cell culture protocol for generating epidermal human skin tissue using primary human keratinocytes, dermal fibroblasts, and collagen-coated transwell inserts.

Related Content

Diagnostic immunohistochemistry overview highlights its importance in cancer diagnosis and detecting infectious diseases in modern clinical pathology.

Diagnostic immunohistochemistry overview highlights its importance in cancer diagnosis and detecting infectious diseases in modern clinical pathology.

Diagnostic immunohistochemistry overview highlights its importance in cancer diagnosis and detecting infectious diseases in modern clinical pathology.

Diagnostic immunohistochemistry overview highlights its importance in cancer diagnosis and detecting infectious diseases in modern clinical pathology.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service