Skip to Content
Merck
All Photos(1)

Key Documents

270A-1

Sigma-Aldrich

IgM Rabbit Polyclonal Antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

100
500

conjugate

unconjugated

antibody form

Ig fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

description

For In Vitro Diagnostic Use in Select Regions (See Chart)

form

buffered aqueous solution

species reactivity

human

packaging

vial of 0.1 mL concentrate (270A-14)
vial of 0.5 mL concentrate (270A-15)
bottle of 1.0 mL predilute (270A-17)
vial of 1.0 mL concentrate (270A-16)
bottle of 7.0 mL predilute (270A-18)

manufacturer/tradename

Cell Marque®

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:100-1:500

control

tonsil

shipped in

wet ice

storage temp.

2-8°C

visualization

cytoplasmic

General description

Anti-IgM antibody reacts with surface immunoglobulin IgM mu chains. IgM is one of the predominant surface immunoglobulins on B-lymphocytes. This antibody is useful when identifying lymphomas, plasmacytomas, and B-cell lineage derived Hodgkin′s lymphomas. Due to the restricted expression of heavy and light chains in these diseases, demonstration of B-cell lymphomas is possible with clonal gene rearrangement studies.

Quality


IVD

IVD

IVD

RUO

Linkage

IgM (polyclonal) Positive Control Slides, Product No. 270S, are available for immunohistochemistry (formalin-fixed, paraffin-embedded sections).

Physical form

Solution in Tris Buffer, pH 7.3-7.7, with 1% BSA and <0.1% Sodium Azide

Preparation Note

Download the IFU specific to your product lot and formatNote: This requires a keycode which can be found on your packaging or product label.

Other Notes

For Technical Service please contact: 800-665-7284 or email: [email protected]

Legal Information

Cell Marque is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C J de Boer et al.
Annals of oncology : official journal of the European Society for Medical Oncology, 8 Suppl 2, 109-117 (1997-01-01)
Mantle-cell lymphoma comprises 2%-10% of all non-Hodgkin's lymphomas (NHLs). Patients present with generalized disease, and have a poor prognosis. Three different histologic patterns (mantle zone, nodular, and diffuse) and three different cytological variants (classical, blastic, and pleomorphic) have been described.
Masaru Kojima et al.
APMIS : acta pathologica, microbiologica, et immunologica Scandinavica, 110(12), 875-880 (2003-03-21)
Nodal marginal zone B-cell lymphomas (NMZBL) occasionally represent prominent plasma cell differentiation. Recently, we presented a patient with NMZBL who exhibited histological features that resembled plasmacytoma arising from a localized plasma cell variant of Castleman's disease. To further clarify the
Tissue section immunologic methods in lymphomas
Warnake, R., et al.
Diagnostic Immunohistochemistry (Masson Publishing), 203-221 (1981)
Manual of Diagnostic Antibodies for Immunohistology, 217-219 (1999)
S E Pambuccian et al.
The American journal of surgical pathology, 21(2), 179-186 (1997-02-01)
Tumoral amyloidosis (amyloidoma) of bone is a rare condition characterized by the massive destructive deposition of AL amyloid in bones. We report three cases. The patients ranged in age from 45 to 78 years and had tumors located in the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service