Skip to Content
Merck
All Photos(1)

Key Documents

26420

Sigma-Aldrich

Chloro(2,2′:6′,2″-terpyridine)platinum(II) chloride dihydrate

≥99.0% (AAS)

Synonym(s):

Dichloro(2,2′:6′,2′′-terpyridine)platinum(II) dihydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H11Cl2N3Pt · 2H2O
CAS Number:
Molecular Weight:
535.28
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.32

Quality Level

Assay

≥99.0% (AAS)

SMILES string

O.O.Cl[Pt]Cl.c1ccc(nc1)-c2cccc(n2)-c3ccccn3

InChI

1S/C15H11N3.2ClH.2H2O.Pt/c1-3-10-16-12(6-1)14-8-5-9-15(18-14)13-7-2-4-11-17-13;;;;;/h1-11H;2*1H;2*1H2;/q;;;;;+2/p-2

InChI key

WEBUYGBVKZYTMT-UHFFFAOYSA-L

Other Notes

An intercalator of helical and double stranded DNA; Spectroscopic tag for proteins. Selective covalent labeling of histidine in cytochromes c; Intercalation

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S. J. Lippard
Accounts of Chemical Research, 11, 211-211 (1987)
K W Jennette et al.
Proceedings of the National Academy of Sciences of the United States of America, 71(10), 3839-3843 (1974-10-01)
The red, cationic complex 2-hydroxy-ethanethiolato(2,2',2''-terpyridine)platinum(II), [(terpy)-Pt(SCH(2)CH(2)OH)](+), binds strongly to DNA by a mechanism involving intercalation. By means of fluorescence spectroscopy, the platinum complex was shown to inhibit competitively the binding of the intercalating dye ethidium bromide to calf thymus DNA.
E.M.A. Ratilla et al.
Journal of the American Chemical Society, 109, 4592-4592 (1987)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service