Skip to Content
Merck
All Photos(1)

Key Documents

259A-1

Sigma-Aldrich

Glucagon Rabbit Polyclonal Antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

100
500

conjugate

unconjugated

antibody form

Ig fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

description

For In Vitro Diagnostic Use in Select Regions (See Chart)

form

buffered aqueous solution

species reactivity

human

packaging

vial of 0.1 mL concentrate (259A-14)
vial of 0.5 mL concentrate (259A-15)
bottle of 1.0 mL predilute (259A-17)
vial of 1.0 mL concentrate (259A-16)
bottle of 7.0 mL predilute (259A-18)

manufacturer/tradename

Cell Marque

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:100-1:500

control

pancreas

shipped in

wet ice

storage temp.

2-8°C

visualization

cytoplasmic

Gene Information

human ... GCG(2641)

General description

The normal islets of Langerhans (pancreas) contain four major cell types, 15-20% of which are alpha cells which produce glucagon. Alpha cell tumors can be divided into two distinct types; those associated with glucagonoma syndrome (necrolytic migratory erythema, diabetes, anemia, stomatitis, weight loss, venous thromboses, diarrhea, and sometimes psychiatric disturbances), and those which are not. Those associated with the syndrome tend to be large and solitary and there is a high incidence of malignancy. Those glucagonomas not associated with the syndrome tend to be multiple and small and are nearly always benign.Immunohistochemically, the two types of glucagonoma are reactive to the glucagon antibody, though occasionally only weakly.

Quality


IVD

IVD

IVD

RUO

Linkage

Glucagon (polyclonal) Positive Control Slides, Product No. 259S, are available for immunohistochemistry (formalin-fixed, paraffin-embedded sections).

Physical form

Solution in Tris Buffer, pH 7.3-7.7, with 1% BSA and <0.1% Sodium Azide

Preparation Note

Download the IFU specific to your product lot and formatNote: This requires a keycode which can be found on your packaging or product label.

Other Notes

For Technical Service please contact: 800-665-7284 or email: service@cellmarque.com

Legal Information

Cell Marque is a trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rosai and Ackerman's Surgical Pathology, 10th ed., 1030-1031 (2011)
Diagnostic Immunohistochemistry: Theranostic and Genomic Applications (2010)
Aixia Sun et al.
Transplantation direct, 10(7), e1658-e1658 (2024-06-17)
Transplantation of human-induced pluripotent stem cell (hiPSC)-derived islet organoids is a promising cell replacement therapy for type 1 diabetes (T1D). It is important to improve the efficacy of islet organoids transplantation by identifying new transplantation sites with high vascularization and
Q A Hamid et al.
Histopathology, 10(2), 119-133 (1986-02-01)
Histological diagnosis of neuroendocrine tumours can be hampered by their lack of peptide or amine immunoreactivity. In order to assess the usefulness of a range of specific and general markers of neuroendocrine differentiation, 10 pancreatic endocrine tumours, associated with high
Yang Xiao et al.
Diabetologia, 66(6), 1142-1155 (2023-03-15)
Glucagon-stimulated hepatic gluconeogenesis contributes to endogenous glucose production during fasting. Recent studies suggest that TGF-β is able to promote hepatic gluconeogenesis in mice. However, the physiological relevance of serum TGF-β levels to human glucose metabolism and the mechanism by which

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service