Skip to Content
Merck
All Photos(1)

Key Documents

21279

Sigma-Aldrich

Calyculin A from Discodermia calyx

≥90% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C50H81N4O15P
CAS Number:
Molecular Weight:
1009.17
Beilstein:
4903216
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥90% (HPLC)

form

solid

storage temp.

−20°C

SMILES string

COC[C@@H]([C@H](O)[C@H](O)C(=O)NCC[C@H](C)c1nc(\C=C\C[C@@H]2O[C@]3(C[C@@H](O)[C@@H]2C)OC([C@H](C[C@H](O)[C@H](C)[C@H](O)[C@H](C)\C=C(C)\C(C)=C\C=C\C(C)=C/C#N)OC)[C@H](OP(O)(O)=O)C3(C)C)co1)N(C)C

InChI

1S/C50H81N4O15P/c1-29(20-22-51)16-14-17-30(2)32(4)24-33(5)42(57)35(7)38(55)25-41(65-13)45-46(69-70(61,62)63)49(8,9)50(68-45)26-39(56)34(6)40(67-50)19-15-18-36-27-66-48(53-36)31(3)21-23-52-47(60)44(59)43(58)37(28-64-12)54(10)11/h14-18,20,24,27,31,33-35,37-46,55-59H,19,21,23,25-26,28H2,1-13H3,(H,52,60)(H2,61,62,63)/b16-14+,18-15+,29-20-,30-17+,32-24+/t31-,33+,34-,35-,37-,38-,39+,40-,41-,42+,43-,44-,45+,46-,50+/m0/s1

InChI key

FKAWLXNLHHIHLA-YCBIHMBMSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Inhibitor of protein phosphatases types 1 and 2A; marine toxin, potent tumor promotor.

Other Notes

Potent inhibitor of protein phosphatases 1 and 2A. More than 10 times more effective for PP1 than okadaic acid

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Skin Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L.C. Starke et al.
The American Journal of Physiology, 264 (1993)
Haruhiko Miyata et al.
Cytoskeleton (Hoboken, N.J.), 69(4), 207-220 (2012-01-27)
Many motile processes are regulated such that movement occurs only upon activation of a signaling cascade. Sperm from a variety of species are initially quiescent and must be activated prior to beating. The signaling events leading to the activation and
Lucas H Ting et al.
American journal of physiology. Heart and circulatory physiology, 302(11), H2220-H2229 (2012-03-27)
Endothelial cells respond to fluid shear stress through mechanotransduction responses that affect their cytoskeleton and cell-cell contacts. Here, endothelial cells were grown as monolayers on arrays of microposts and exposed to laminar or disturbed flow to examine the relationship among
A V Mel'nitskaia et al.
Tsitologiia, 54(2), 143-148 (2012-05-18)
Using voltage-clamp technique, the possible role of the cytoskeleton in the effect of pharmacological analogue of oxidized glutathione (GSSG), drug glutoxim, on Na+ transport in the frog Rana temporaria skin was investigated. It was shown for the first time that
Michael A Kuefner et al.
International journal of radiation biology, 89(6), 424-432 (2013-02-01)
The purpose of this study was to investigate the effect of calyculin A on the number of γ-H2AX foci (phosphorylated histone variant 2AX) in lymphocytes after in vitro and in vivo irradiation with rather low doses as they are used

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service