Skip to Content
Merck
All Photos(1)

Key Documents

205R-1

Sigma-Aldrich

CD5 (SP19) Rabbit Monoclonal Antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

100
500

conjugate

unconjugated

antibody form

culture supernatant

antibody product type

primary antibodies

clone

SP19, monoclonal

description

For In Vitro Diagnostic Use in Select Regions (See Chart)

form

buffered aqueous solution

species reactivity

human

packaging

vial of 0.1 mL concentrate (205R-14)
vial of 0.5 mL concentrate (205R-15)
bottle of 1.0 mL predilute (205R-17)
vial of 1.0 mL concentrate (205R-16)
bottle of 7.0 mL predilute (205R-18)

manufacturer/tradename

Cell Marque

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:100-1:500

isotype

IgG

control

tonsil

shipped in

wet ice

storage temp.

2-8°C

visualization

membranous

Gene Information

human ... CD5(921)

General description

Anti-CD5 is a pan T-cell marker that also reacts with a range of neoplastic B-cells, e.g. chronic lymphocytic leukemia/small lymphocytic lymphoma (CLL/SLL), mantle cell lymphoma, and a subset (~10%) of diffuse large B-cell lymphoma. CD5 aberrant expression is useful in making a diagnosis of mature T-cell neoplasms. Anti-CD5 detection is diagnostic in CLL/SLL within a panel of other B-cell markers, especially one that includes anti-CD23. Anti-CD5 is also very useful in differentiating among mature small lymphoid cell malignancies. In addition, anti-CD5 can be used in distinguishing thymic carcinoma (+) from thymoma (-), especially when combined with CD117 and CK7. Anti-CD5 does not react with granulocytes or monocytes.

Quality


IVD

IVD

IVD

RUO

Linkage

CD5 Positive Control Slides, Product No. 205S, are available for immunohistochemistry (formalin-fixed, paraffin-embedded sections).

Physical form

Solution in Tris Buffer, pH 7.3-7.7, with 1% BSA and <0.1% Sodium Azide

Preparation Note

Download the IFU specific to your product lot and formatNote: This requires a keycode which can be found on your packaging or product label.

Other Notes

For Technical Service please contact: 800-665-7284 or email: service@cellmarque.com

Legal Information

Cell Marque is a trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chung-Che Chang et al.
Modern pathology : an official journal of the United States and Canadian Academy of Pathology, Inc, 15(10), 1051-1057 (2002-10-16)
CD5 expression in neoplastic large B-cells in T-cell/histiocyte-rich large B-cell lymphoma has not been reported, to the best of our knowledge. Here we describe the first case of CD5+ T-cell/histiocyte-rich large B-cell lymphoma that is well documented by histomorphology, immunohistochemistry
CD5+ B lymphocytes.
M T Kasaian et al.
Proceedings of the Society for Experimental Biology and Medicine. Society for Experimental Biology and Medicine (New York, N.Y.), 197(3), 226-241 (1991-07-01)
Ben Hatano et al.
Pathology international, 52(5-6), 400-405 (2002-07-09)
Peripheral T-cell lymphomas (PTCL) with nodular growth patterns are very rare, with only 17 cases reported previously. Here, we report a case of PTCL with a nodular growth pattern. The patient was an 81-year-old Japanese woman who complained of malaise
J K Chan et al.
Histopathology, 25(6), 517-536 (1994-12-01)
A new classification of lymphoid neoplasms, mostly based on existing terminology, is proposed by the International Lymphoma Study Group. The proposed classification was reached through a consensus of the members, despite their diverse backgrounds, and consists of a listing of
N H Jones et al.
Nature, 323(6086), 346-349 (1986-09-01)
The T1/Leu-1/CD5 molecule, a human T-cell surface glycoprotein of relative molecular mass (Mr) 67,000, has been implicated in the proliferative response of activated T cells and in T-cell helper function. A similar involvement in T-cell proliferation has been reported for

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service