Skip to Content
Merck
All Photos(1)

Key Documents

16702

Sigma-Aldrich

(R)-Pantetheine

≥95.0% (HPLC)

Synonym(s):

(2R)-2,4-Dihydroxy-N-[3-[(2-mercaptoethyl)amino]-3-oxopropyl]-3,3-dimethylbutanamide, (R)-2,4-Dihydroxy-N-[2-[(2-mercaptoethyl)carbamoyl]ethyl]-3,3-dimethylbutyramide, (D)-(+)-Pantetheine, N-(Pantothenyl)-β-aminoethanethiol, LBF, Lactobacillus bulgaricus factor

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H22N2O4S
CAS Number:
Molecular Weight:
278.37
Beilstein:
1714196
MDL number:
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.79

Assay

≥95.0% (HPLC)

form

solid

optical activity

[α]/D 17±2°, c = 1 in H2O

color

white

storage temp.

−20°C

SMILES string

OCC(C)(C)[C@@H](O)C(NCCC(NCCS)=O)=O

InChI

1S/C11H22N2O4S/c1-11(2,7-14)9(16)10(17)13-4-3-8(15)12-5-6-18/h9,14,16,18H,3-7H2,1-2H3,(H,12,15)(H,13,17)/t9-/m0/s1

InChI key

ZNXZGRMVNNHPCA-VIFPVBQESA-N

Application


  • Structures of carboxylic acid reductase reveal domain dynamics underlying catalysis: This study investigates the structural and dynamic aspects of carboxylic acid reductase enzymes, which are relevant for the conversion processes involving (R)-pantetheine and related compounds (Gahloth et al., 2017).

Biochem/physiol Actions

Metabolite in carbapenem biosynthesis, pantothenate and CoA biosynthesis and biosynthesis of secondary metabolites.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P Dykes et al.
Journal of wound care, 26(9), 552-557 (2017-09-08)
Preventing moisture damage and breakdown of the skin can be a particular challenge for patients with incontinence. The level of protection offered by various skin protectant products can vary according to the chemical nature of the formulation and can decrease
Lindsay E O'Donnell et al.
Journal of medical microbiology, 66(1), 54-60 (2016-12-30)
Approximately 20  % of the UK population wear some form of denture prosthesis, resulting in denture stomatitis in half of these individuals. Candida albicans is primarily attributed as the causative agent, due to its biofilm -forming ability. Recently, there has
Haeju Jo et al.
Materials (Basel, Switzerland), 13(18) (2020-09-25)
This study investigated the mechanical properties of steel in flanges, with the goal of obtaining high strength and high toughness. Quenching was applied alone or in combination with tempering at one of nine combinations of three temperatures TTEM and durations
Matthew F Crum et al.
Molecular pharmaceutics, 14(2), 394-405 (2016-12-22)
The current studies sought to explore the impact of drug supersaturation and precipitation during the dispersion and digestion of lipid-based formulations (LBFs), on in vivo absorption using a coupled in vitro digestion-in vivo perfusion absorption model. Fenofibrate absorption was evaluated
Jun Yang et al.
Applied optics, 57(10), 2490-2498 (2018-05-02)
On-site measurements and defect detection are of great importance for precision ground steel rollers due to their large dimension and weight. In addition to dimensional error, form accuracy, surface roughness, and surface/sub-surface cracks, there also exist optical defect requirements for

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service