Skip to Content
Merck
All Photos(1)

Key Documents

135R-1

Sigma-Aldrich

CD35 (EP197) Rabbit Monoclonal Primary Antibody

Synonym(s):

C3-binding protein, C3BR, C3b/C4b receptor, C4BR, CD35 antigen, CR1, Complement Receptor 1, Complement component (3b/4b) receptor 1 (Knops blood group), Knops blood group antigen

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

100
500

conjugate

unconjugated

antibody form

culture supernatant

antibody product type

primary antibodies

clone

EP197, monoclonal

description

For In Vitro Diagnostic Use in Select Regions (See Chart)

form

buffered aqueous solution

species reactivity

human

packaging

vial of 0.1 mL concentrate (135R-14)
vial of 0.1 mL concentrate Research Use Only (135R-14-RUO)
vial of 0.5 mL concentrate (135R-15)
vial of 1.0 mL concentrate (135R-16)
vial of 1.0 mL concentrate Research Use Only (135R-16-RUO)
vial of 1.0 mL pre-dilute Research Use Only (135R-17-RUO)
vial of 1.0 mL predilute ready-to-use (135R-17)
vial of 7.0 mL pre-dilute ready-to-use Research Use Only (135R-18-RUO)
vial of 7.0 mL predilute ready-to-use (135R-18)

manufacturer/tradename

Cell Marque

concentration

0.1-5.0 μg/mL (predilute)
2.5-125 μg/mL (concentrate)

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:25-1:100 (concentrated)

isotype

IgG

control

tonsil

shipped in

wet ice

storage temp.

2-8°C

visualization

membranous

Gene Information

human ... CR1(1378)

General description

CD35 (complement receptor 1) is a cell membrane-bound, monomeric glycoprotein found on numerous cell types including erythrocytes, leukocytes, glomerular podocytes, and follicular dendritic cells. The primary function of CD35 is to serve as the cellular receptor for C3b and C4b, the most important components of the complement system leading to clearance of foreign macromolecules. The Knops blood group system is a system of antigens located on this protein. The protein mediates cellular binding to particles and immune complexes that have activated complement. Follicular dendritic cells (FDC) are restricted to the B-cell regions of secondary lymphoid follicles. CD35 antigen is found on erythrocytes, B cells, a subset of T cells, monocytes, as well as eosinophils and neutrophils. Anti-CD35 is considered a mature B-cell marker which labels follicular dendritic reticulum cells and tumors derived from such cells such as follicular dendritic cell tumor/sarcoma.

Quality

United States - IVD
Canada - IVD
European Union - IVD
Japan - RUO

Linkage

CD35 Positive Control Slides, Product No. 135S, are available for immunohistochemistry (formalin-fixed, paraffin-embedded sections).

Physical form

Solution in Tris Buffer, pH 7.3-7.7, with 1% BSA and <0.1% Sodium Azide

Preparation Note

Download the IFU specific to your product lot and format
Note: This requires a keycode which can be found on your packaging or product label.

Download the latest released IFU
Note: This IFU may not apply to your specific product lot.

Other Notes

For Technical Service please contact: 800-665-7284 or email: service@cellmarque.com

Legal Information

Cell Marque is a trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tumours of histiocytes and accessory dendritic cells: an immunohistochemical approach to classification from the International Lymphoma Study Group based on 61 cases.
Pileri SA, Grogan TM, Harris NL, et al.
Histopathology, 41(1), 1-29 (2002)
Immunohistochemical recognition of human follicular dendritic cell (FDCs) in routinely processed paraffin section.
Kunihikom, M., et al.
The Journal of Histochemistry and Cytochemistry, 50, 1475-1486 (2002)
Extranodal follicular dendritic cell sarcoma of the head and neck region: three new cases, with a review of the literature.
Biddle DA, Ro JY, Yoon GS, et al.
Modern Pathology, 15(1), 50-58 (2002)
Germinal centre-like versus undifferentiated stromal immunophenotypes in follicular lymphoma.
Chang KC, Huang X, Medeiros LJ, et al.
The Journal of Pathology, 201(3), 404-412 (2003)
Mediastinal mixed dendritic cell sarcoma with hybrid features.
Dillon KM, Hill CM, Cameron CH, et al.
Journal of Clinical Pathology, 55(10), 791-794 (2002)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service