Skip to Content
Merck

108315

SAFC

TRIS hydrochloride

EMPROVE® EVOLVE

Pharma Manufacturing

Synonym(s):

Trizma® hydrochloride, TRIS HCl, TRIS hydrochloride, Tris(hydroxymethyl)aminomethane hydrochloride, Tromethane hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2C(CH2OH)3 · HCl
CAS Number:
Molecular Weight:
157.60
Beilstein:
3675235
MDL number:
UNSPSC Code:
12161700
Pricing and availability is not currently available.

biological source

synthetic

Quality Level

description

Manufactured under appropriate controls for use as a raw material in BioProcessing

product line

EMPROVE® EVOLVE

form

powder

technique(s)

cell culture | mammalian: suitable

impurities

Endotoxin, tested
Microbial, tested
Trace Metals, tested

useful pH range

7.0-9.0

pKa (25 °C)

8.1

suitability

suitable for manufacturing use

foreign activity

Cytotoxicity, contains (tested)
DNase, contains (tested)
NICKase, contains (tested)
Protease, contains (tested)
RNase, contains (tested)

SMILES string

Cl.NC(CO)(CO)CO

InChI

1S/C4H11NO3.ClH/c5-4(1-6,2-7)3-8;/h6-8H,1-3,5H2;1H

InChI key

QKNYBSVHEMOAJP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Our SAFC® portfolio of high-quality raw materials for use in biopharmaceutical processing withstands strict quality control procedures plus the documentation and expertise to help our customers meet requirements as defined by the M-Clarity Program.

M-Clarity Program

As part of our EMPROVE® Program, our raw materials are offered with EMPROVE® Dossiers which provide comprehensive, up-to-date documentation to help you navigate regulatory challenges, manage risks, and improve your manufacturing processes.

Buffer quality is vital for the success of biopharmaceutical processes, because buffers are indispensable in nearly every production step.

Our broad portfolio of buffer materials manufactured under appropriate controls is tailored to your needs. Ranging from non-GMP grades for low-risk application, to IPEC-PQG GMP for higher-risk applications, we have products covering all your manufacturing needs.

Application

Tris HCl is a commonly used biological buffer. Tris/Tris HCl as a buffering system has a very broad usage in biological systems for biopharmaceutical manufacturing, cell culture, diagnostics and molecular biology. The pKa of Tris is 7.77 which corresponds closely to a physiological pH of 7.36.

Tris HCl is used as the conjugate buffer component with Tris/Tromethamine/Trizma in biopharmaceutical buffer formulations (both upstream and downstream applications), electrophoresis separations and diagnostic reagent formulations.

Other Notes

The pH values of all buffers are temperature and concentration dependent. For Tris buffers, pH increases about 0.03 unit per °C decrease in temperature, and decreases 0.03-0.05 unit per ten-fold dilution. For precise applications, use a carefully calibrated pH meter with a glass/calomel combination electrode.

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany
SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany
Trizma is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


What is the Emprove® Program?

The Emprove® Program is a system providing comprehensive and thorough documentation of our filters and single-use components, pharma raw materials, and starting materials. Four document types are included with an Emprove® Program subscription:

Material Qualification Dossier

Free of charge - Sign in to download

Material Qualification Dossier

Free of charge - Sign in to download

Quality Management Dossier (Open Access)

Free of charge - Sign in to download

Quality Management Dossier (Open Access)

Free of charge - Sign in to download

Operational Excellence Dossier

Available by subscription or for a fee

Learn more about the benefits an Emprove® Program subscription can provide
Why are these documents unavailable?
Emprove® Dossiers contain information intended to support the registration process and risk assessment activities of pharmaceutical producers and contain confidential information. Please contact us if you would like access.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Difficulty Finding Your Product Or Lot/Batch Number?

Product numbers are combined with Pack Sizes/Quantity when displayed on the website (example: T1503-25G). Please make sure you enter ONLY the product number in the Product Number field (example: T1503).

Example:

T1503
Product Number
-
25G
Pack Size/Quantity

Additional examples:

705578-5MG-PW

PL860-CGA/SHF-1EA

MMYOMAG-74K-13

1000309185

enter as 1.000309185)

Having trouble? Feel free to contact Technical Service for assistance.

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service