Skip to Content
Merck
All Photos(1)

Documents

10791

Sigma-Aldrich

Antipain

>50000 U/mg

Synonym(s):

[(S)-1-Carboxy-2-phenylethyl]carbamoyl-L-arginyl-L-valyl-argininal

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H44N10O6
CAS Number:
Molecular Weight:
604.70
Beilstein:
6837629
EC Number:
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

Quality Level

specific activity

>50000 U/mg

Mode of action

enzyme | inhibits

storage temp.

−20°C

SMILES string

[H]C(=O)C(CCCNC(N)=N)NC(=O)[C@@H](NC(=O)[C@H](CCCNC(N)=N)NC(=O)N[C@@H](Cc1ccccc1)C(O)=O)C(C)C

InChI

1S/C27H44N10O6/c1-16(2)21(23(40)34-18(15-38)10-6-12-32-25(28)29)37-22(39)19(11-7-13-33-26(30)31)35-27(43)36-20(24(41)42)14-17-8-4-3-5-9-17/h3-5,8-9,15-16,18-21H,6-7,10-14H2,1-2H3,(H,34,40)(H,37,39)(H,41,42)(H4,28,29,32)(H4,30,31,33)(H2,35,36,43)/t18?,19-,20-,21-/m0/s1

InChI key

SDNYTAYICBFYFH-KTYMLHDXSA-N

General description

Chemical structure: peptide

Biochem/physiol Actions

Reversible inhibitor of serine/cysteine proteases and some trypsin-like serine proteases. Its action resembles leupeptin; however, its plasmin inhibition is less and its cathepsin A inhibition is more than that observed with leupeptin. Chronic administration can reduce the frequency of chemically induced transformation in BALB/c-/3T3 cells.
Inhibits serine/cysteine proteases and some trypsin-like proteases. Compare activity to that of leupeptin. Used to evaluate the role of proteases in cell transformations. Used to help identify new proteases.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Unit Definition

1 U corresponds to the amount of inhibitor which reduces the trypsin activity by 1 BAEE-U. (1 BAEE-U is the amount of enzyme which increases the absorbance at 253 nm by 0.001 per minute at pH 7.6 and 25°C; BAEE, Cat. No. 12880, as substrate)

Other Notes

Protease inhibitor

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Raquel Alves Dos Santos et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 46(2), 671-677 (2007-10-27)
The use of antioxidants during chemotherapy has been shown to reduce or prevent the undesirable effects experienced by healthy cells. Micronutrient selenium is well known for its antioxidant properties; however, selenium exhibits a bimodal nature in that both its beneficial
Tanya V Kogan et al.
The Journal of infectious diseases, 189(11), 1965-1973 (2004-05-15)
Aspergillus fumigatus is an opportunistic pathogenic fungus that predominantly infects the respiratory system. Penetration of the lung alveolar epithelium is a key step in the infectious process. The cytoskeleton of alveolar epithelial cells forms the cellular basis for the formation
André L S Santos et al.
International journal for parasitology, 36(1), 47-56 (2005-11-29)
In this study, we report the ultrastructural and growth alterations caused by cysteine peptidase inhibitors on the plant trypanosomatid Phytomonas serpens. We showed that the cysteine peptidase inhibitors at 10 microM were able to arrest cellular growth as well as
Catherine Gaitanaki et al.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology, 13(3), 173-180 (2003-07-24)
To examine whether the calpain-calpastatin system is activated during the calcium paradox in the isolated perfused pigeon heart, we separated the protease from its inhibitor calpastatin and studied its kinetic properties. The protease exhibits kinetic properties similar to those of
E Isogai et al.
Mutation research, 403(1-2), 215-222 (1998-09-03)
In order to examine the relationship between activation of an antipain-sensitive protease and suppression of mutability in UV (UVC)-irradiated human cells, a human cell variant with the high protease activity induced by UV was established and characterized for its susceptibility

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service