Skip to Content
Merck
All Photos(1)

Key Documents

10050

Sigma-Aldrich

Amygdalin

BioXtra, ≥97.0% (HPLC)

Synonym(s):

D-Mandelonitrile 6-O-β-D-glucosido-β-D-glucoside, D-Mandelonitrile-β-gentiobioside, Amygdaloside, Laetrile

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H27NO11
CAS Number:
Molecular Weight:
457.43
Beilstein:
66856
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

biological source

synthetic

Quality Level

product line

BioXtra

Assay

≥97.0% (HPLC)

form

powder

optical activity

[α]20/D −39±2°, c = 2% in H2O

technique(s)

HPLC: suitable

loss

≤7.5% loss on drying, 110 °C

color

white to off-white

solubility

H2O: 0.1 g/mL, clear to very faintly turbid, colorless (hot)

anion traces

chloride (Cl-): ≤50 mg/kg
sulfate (SO42-): ≤300 mg/kg

cation traces

Ca: ≤30 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤300 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Na: ≤50 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Zn: ≤5 mg/kg

SMILES string

OC[C@H]1O[C@@H](OC[C@H]2O[C@@H](OC(C#N)c3ccccc3)[C@H](O)[C@@H](O)[C@@H]2O)[C@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C20H27NO11/c21-6-10(9-4-2-1-3-5-9)30-20-18(28)16(26)14(24)12(32-20)8-29-19-17(27)15(25)13(23)11(7-22)31-19/h1-5,10-20,22-28H,7-8H2/t10?,11-,12-,13-,14-,15+,16+,17-,18-,19-,20-/m1/s1

InChI key

XUCIJNAGGSZNQT-SWRVSKMJSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Amygdalin, a cyanide containing glycoside, may be used as a substrate to identify, differentiate and characterize enzymes such as maltase(s), emulsin(s) and β-glucosidase(s).

Biochem/physiol Actions

Cyanogenic glycoside that is a component of bitter almonds and apricot pits. There is no scientific evidence that amygdalin itself is an effective anti-cancer agent. Recent studies using β−glucoside linked to a tumor-associated monoclonal antibody to release cyanide at the tumor cell has shown significant cytotoxicity.

Other Notes

To gain a comprehensive understanding of our extensive range of Disaccharides for your research, we encourage you to visit our Carbohydrates Category page.
Review: Cyanogenic glycosides

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E.E. Conn
Int. Rev. Biochem. (Biochemistry of Nutrition 1A, A. Neuberger, T.H. Jukes, eds., 27, 21-21 (1979)
Alexander M Abdelnoor
Expert opinion on therapeutic patents, 19(8), 1057-1071 (2009-07-02)
It is hypothesized that psoriasis is an autoimmune disease. The most recent therapeutic approach that proved to be more effective than earlier methods of treatment is the use of mAb/fusion proteins. Efforts nowadays are focused on investigating the antipsoriatic affect
Ebenezer O Farombi et al.
International journal of environmental research and public health, 8(6), 2533-2555 (2011-07-22)
Recently, considerable attention has been focused on dietary and medicinal phytochemicals that inhibit, reverse or retard diseases caused by oxidative and inflammatory processes. Vernonia amygdalina is a perennial herb belonging to the Asteraceae family. Extracts of the plant have been
Lishuang Yu et al.
Electrophoresis, 32(2), 218-222 (2011-01-22)
A new method for separation and determination of amygdalin and its epimer (neoamygdalin) in the epimerization of amygdalin by MEEKC is proposed. For the chiral separation of amygdalin and neoamygdalin, a running buffer composed of 80 mM sodium cholate, 5.0% v/v
Geraldine A Wright et al.
Current biology : CB, 20(24), 2234-2240 (2010-12-07)
Avoiding toxins in food is as important as obtaining nutrition. Conditioned food aversions have been studied in animals as diverse as nematodes and humans [1, 2], but the neural signaling mechanisms underlying this form of learning have been difficult to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service