Skip to Content
Merck
All Photos(1)

Key Documents

09978

Sigma-Aldrich

Ammonium sulfate

BioUltra, ≥99.0% (T)

Synonym(s):

Mascagnite

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(NH4)2SO4
CAS Number:
Molecular Weight:
132.14
EC Number:
MDL number:
UNSPSC Code:
12352314
eCl@ss:
38030408
PubChem Substance ID:
NACRES:
NA.26
Pricing and availability is not currently available.

product line

BioUltra

Quality Level

Assay

≥99.0% (T)

form

crystals

impurities

insoluble matter, passes filter test

ign. residue

≤0.01% (as SO4)

loss

≤0.1% loss on drying, 110 °C

pH

5.0-6.0 (25 °C, 1 M in H2O)

mp

>280 °C (dec.) (lit.)

solubility

H2O: 1 M at 20 °C, clear, colorless

density

1.77 g/mL at 25 °C (lit.)

anion traces

chloride (Cl-): ≤5 mg/kg
nitrate (NO3-): ≤50 mg/kg
phosphate (PO43-): ≤5 mg/kg

cation traces

Al: ≤5 mg/kg
As: ≤0.2 mg/kg
Ba: ≤5 mg/kg
Bi: ≤5 mg/kg
Ca: ≤10 mg/kg
Cd: ≤1 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤2 mg/kg
Fe: ≤2 mg/kg
K: ≤50 mg/kg
Li: ≤5 mg/kg
Mg: ≤50 mg/kg
Mn: ≤5 mg/kg
Mo: ≤5 mg/kg
Na: ≤50 mg/kg
Ni: ≤5 mg/kg
Pb: ≤2 mg/kg
Sr: ≤5 mg/kg
Zn: ≤1 mg/kg

λ

1 M in H2O

UV absorption

λ: 260 nm Amax: ≤0.037
λ: 280 nm Amax: ≤0.030

SMILES string

N.N.OS(O)(=O)=O
N.N.OS(O)(=O)=O

InChI

1S/2H3N.H2O4S/c;;1-5(2,3)4/h2*1H3;(H2,1,2,3,4)

InChI key

BFNBIHQBYMNNAN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ammonium sulfate is naturally present as a rare mineral mascagnite in volcanic fumaroles and due to coal fires in some dumps.

Application

Ammonium sulfate has been used:
  • to prepare mobile phase A for hydrophobic interaction chromatography (HIC)
  • as reference material for the thermal decomposition of NH4NO3 (S1–S6) thermal decomposition to N2O (S1-N2O–S6-N2O)
  • to prepare the stock solution for M63 medium
  • as a reagent in molecular biology and chromatography
  • in the precipitation and fractionation of proteins
  • in the purification of antibodies
  • for the crystallographic analysis of nucleic acids and proteins
  • in high-performance liquid chromatography (HPLC) analysis of proteins, such as in hydrophobic interaction chromatography
  • for Sepharose chromatography of tRNAs using reverse salt gradients
  • in long PCR buffer, in PCR lysis solution, and second-strand cDNA synthesis
  • HPLC analysis of phospholipids

Biochem/physiol Actions

Ammonium sulfate is an inorganic salt, which is mostly used as a soil fertilizer. It is also used as a flame retardant since it elevates the combustion temperature of the material and lowers maximum weight loss rates. This leads to an upsurge in the production of residue or char.

Other Notes

The most commonly used salt for the fractional precipitation of proteins due to its solubility and effectiveness[1]; Protein crystallization[2]; Sepharose chromatography of tRNAs using reverse salt gradients[3].

Legal Information

Sepharose is a trademark of Cytiva

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sepharose chromatography of transfer ribonucleic acids using reverse salt gradients.
G W Hatfield
Methods in enzymology, 59, 215-218 (1979-01-01)
T.D. Pollard
Methods in Enzymology, 85, 334-334 (1982)
Protein crystallization: the growth of large-scale single crystals.
G L Gilliland et al.
Methods in enzymology, 104, 370-381 (1984-01-01)
Simon Gareth Broadley et al.
Acta crystallographica. Section D, Biological crystallography, 68(Pt 11), 1450-1459 (2012-10-24)
MshB, a zinc-based deacetylase, catalyses a step in the mycothiol biosynthetic pathway that involves the deacetylation of 1-O-(2-acetamido-2-deoxy-α-D-glucopyranosyl)-D-myo-inositol (GlcNAc-Ins), via cleavage of an amide bond, to 1-O-(2-amino-2-deoxy-α-D-glucopyranosyl)-D-myo-inositol (GlcN-Ins) and acetate. In this study, MshB was expressed, purified and crystallized. A
James F Hunter et al.
Environmental science & technology, 48(17), 10227-10234 (2014-08-06)
A large number of organic species emitted into the atmosphere contain cycloalkyl groups. While cyclic species are believed to be important secondary organic aerosol (SOA) precursors, the specific role of cyclic moieties (particularly for species with multiple or fused rings)

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service