Skip to Content
Merck
All Photos(1)

Key Documents

09154

Sigma-Aldrich

L-Xylonic acid lithium salt

≥95.0% (TLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H10O6 · xLi+
CAS Number:
Molecular Weight:
166.13 (free acid basis)
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.25

Quality Level

Assay

≥95.0% (TLC)

form

solid

optical activity

[α]/D -18±2°, c = 1 in H2O

color

white

storage temp.

2-8°C

SMILES string

OC[C@H](O)[C@@H](O)[C@H](O)C(O)=O
OC[C@H](O)[C@@H](O)[C@H](O)C(O)=O

InChI

1S/C5H10O6/c6-1-2(7)3(8)4(9)5(10)11/h2-4,6-9H,1H2,(H,10,11)/t2-,3+,4-/m0/s1

InChI key

QXKAIJAYHKCRRA-NUNKFHFFSA-N

Biochem/physiol Actions

Metabolite of the pentose and glucuronate interconversions and the ascorbate and aldarate metabolism.

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rocio García-Villalba et al.
Journal of chromatography. A, 1195(1-2), 164-173 (2008-05-30)
In this work, capillary electrophoresis-time-of-flight mass spectrometry (CE-TOF-MS) is proposed to identify and quantify the main metabolites found in transgenic soybean and its corresponding non-transgenic parental line both grown under identical conditions. The procedure includes optimization of metabolites extraction, separation
Amanda J Lloyd et al.
The American journal of clinical nutrition, 94(4), 981-991 (2011-08-26)
The lack of robust biological markers of dietary exposure hinders the quantitative understanding of causal relations between diet and health. We aimed to develop an efficient procedure to discover metabolites in urine that may have future potential as biomarkers of
George Cooper et al.
Journal of chromatography. A, 1216(40), 6838-6843 (2009-09-01)
Analysis of compounds in meteorites revealed a need to simultaneously characterize multiple enantiomers of sugar acids (aldonic acids) present in trace amounts. Analyses by gas chromatography-mass spectrometry demonstrated that all but two of the three-carbon through six-carbon straight-chained sugar acid
Wen Shan Yew et al.
Biochemistry, 45(49), 14582-14597 (2006-12-06)
Many members of the mechanistically diverse enolase superfamily have unknown functions. In this report we use both genome (operon) context and screening of a library of acid sugars to assign the L-fuconate dehydratase (FucD) function to a member of the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service