Skip to Content
Merck
All Photos(1)

Key Documents

08061901

U-373 MG (Uppsala) cell line

Synonym(s):

U-373MG Cells, U373-MG Cells, U373MG Cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

biological source

human brain

form

liquid

growth mode

Adherent

karyotype

2n = 46

morphology

Pleomorphic/astrocytoid

receptors

PDGFR alpha and EGFR. See Nistér M. et al., (1988) & (1991)

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human glioblastoma astrocytoma

Cell Line Description

A human glioblastoma astrocytoma derived from a malignant tumour by explant technique. This catalogue entry represents a new deposit of U-373 MG obtained from the originator′s laboratory in Uppsala; it has been designated U-373 MG (Uppsala) to distinguish it from U-373 MG (Sigma Catalogue number 89081403) which is now known to be identical, by Short Tandem Repeat (STR)-PCR profiling, to another glioblastoma cell line, U-251 MG (please read below for more information about this discovery). U-373 MG (Uppsala) has been mycoplasma eradicated at ECACC and the stocks available for supply have undergone a further 10 passages without detection of mycoplasma.
Background to the identity query for the cell line U-373 MG: The American Type Culture Collection (ATCC) reported that their stock of U-373 MG had been shown to have differing genetic properties to stock from the originator′s laboratory, and to share similarities with another glioblastoma cell line, U-251. In light of this, ECACC undertook an investigation into the authenticity of its own stock of the U-373 MG cell line. ECACC found similar results to the ATCC i.e. the stock held as U-373 MG was found to be identical by STR-PCR profiling to U-251. The U-373 MG cell line listed under catalogue number 89081403 has been re-named as ′U-251 (formerly known as U-373 MG)′.

Application

Culture Medium: EMEM (EBSS) + 2mM Glutamine + 1% Non Essential Amino Acids (NEAA) + 1mM Sodium Pyruvate + 10% Fetal Bovine Serum (FBS).

Biochem/physiol Actions

U-373 MG (Uppsala) is a human glioblastoma astrocytoma derived from a malignant tumour by explant technique.

DNA Profile

STR-PCR Data: Amelogenin: X
CSF1PO: 11,12
D13S317: 8
D16S539: 12,13
D5S818: 12
D7S820: 8.2,12
THO1: 7,9.3
TPOX: 8
vWA: 17,18

Culture Medium

EMEM (EBSS) + 2mM Glutamine + 1% Non Essential Amino Acids (NEAA) + 1mM Sodium Pyruvate (NaP) + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 to 1:6 i.e. seeding at 2-4x10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
Cultures from HPA Culture Collections and supplied by Sigma are for research purposes only. Enquiries regarding the commercial use of a cell line are referred to the depositor of the cell line. Some cell lines have additional special release conditions such as the requirement for a material transfer agreement to be completed by the potential recipient prior to the supply of the cell line. Please view the Terms & Conditions of Supply for more information.

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Liping Fang et al.
International journal of molecular medicine, 38(1), 172-182 (2016-06-02)
Previous studies have suggested that jumonji AT-rich interactive domain 1B (JARID1B) plays an important role in the genesis of some types of cancer, and it is therefore considered to be an important drug target protein. Although the expression of JARID1B has been researched
Yosr BenRedjem Romdhane et al.
BioMed research international, 2016, 5318176-5318176 (2016-09-07)
Marine sponges of the Irciniidae family contain both bioactive furanosesterterpene tetronic acids (FTAs) and prenylated hydroquinones (PHQs). Both classes of compounds are known for their anti-inflammatory, antioxidant, and antimicrobial properties and known to display growth inhibitory effects against various human
Mark A Lobas et al.
Nature communications, 10(1), 711-711 (2019-02-14)
Adenosine 5' triphosphate (ATP) is a universal intracellular energy source and an evolutionarily ancient, ubiquitous extracellular signal in diverse species. Here, we report the generation and characterization of single-wavelength genetically encoded fluorescent sensors (iATPSnFRs) for imaging extracellular and cytosolic ATP
Alexander V Aksenov et al.
Journal of medicinal chemistry, 58(5), 2206-2220 (2015-02-12)
Many types of tumor, including glioma, melanoma, non-small cell lung, esophageal, and head and neck cancer, among others, are intrinsically resistant to apoptosis induction and poorly responsive to current therapies with proapoptotic agents. In addition, tumors often develop multidrug resistance
Laurent Jacob et al.
Oncotarget, 7(36), 57851-57865 (2016-08-11)
The neuropilin-plexin receptor complex regulates tumor cell migration and proliferation and thus is an interesting therapeutic target. High expression of neuropilin-1 is indeed associated with a bad prognosis in glioma patients. Q-RTPCR and tissue-array analyses showed here that Plexin-A1 is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service