Skip to Content
Merck
All Photos(2)

Key Documents

R4126

Sigma-Aldrich

Reinecke salt

Synonym(s):

Ammonium reineckate, Ammonium tetrarhodanatodiamminechromate(III), Ammonium tetrathiocyanatodiamminechromate(III), Ammonium tetrathiocyanodiammonochromate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH4[Cr(NH3)2(SCN)4]
CAS Number:
Molecular Weight:
336.43
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21
form:
powder

form

powder

Quality Level

mp

268-272 °C (dec.) (lit.)

SMILES string

N.N.[NH4+].N#CS[Cr-](SC#N)(SC#N)SC#N

InChI

1S/4CHNS.Cr.3H3N/c4*2-1-3;;;;/h4*3H;;3*1H3/q;;;;+3;;;/p-3

InChI key

ZGLIQORZYPZFPW-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Application

Reinecke salt can be used as a precipitation reagent for the precipitation of basic compounds such as amines, amino acids, and alkaloids as their reineckate complexes. It is also used in the spectrophotometric determination of promazine along with alizarin S by the formation of the promazine ion-association complex.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Preparation of histones by the use of Reinecke salt.
Lindh NO and Brantmark BL.
Analytical Biochemistry, 10(3), 415-420 (1965)
M Eisman et al.
Journal of pharmaceutical and biomedical analysis, 11(4-5), 301-305 (1993-04-01)
An automatic method for the determination of amylocaine and bromhexine hydrochloride by atomic absorption spectrometry (AAS) is proposed. The drugs were determined indirectly by formation of reineckates, extraction into 1,2-dichloroethane and measurement of chromium in the organic phase. The chemical
L Campanella et al.
Journal of pharmaceutical and biomedical analysis, 14(8-10), 1047-1054 (1996-06-01)
A new cocaine-sensitive ISFET device based on a cocaine-reineckate ion-pair complex dispersed in a PVC--sebacate matrix has been fabricated. this sensor displays a linear range for cocaine hydrochloride between about 3 x 10(-6) and 2 x 10(-2) M and a
Sheikha M Al-Ghannam
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 69(4), 1188-1194 (2007-08-11)
Three accurate, rapid and simple atomic absorption spectrometric (AAS), conductometric and colorimetric methods were developed for the determination of gatifloxacin (GTF), moxifloxacin (MXF) and sparfloxacin (SPF). The proposed methods depend upon the reaction of ammonium reineckate with the studied drugs
M F el-Shahat et al.
Journal of chemical technology and biotechnology (Oxford, Oxfordshire : 1986), 54(2), 175-181 (1992-01-01)
A spectrophotometric method for the determination of some alkaloids (namely ephedrine HCl, cinchonine HCl, chlorpheniramine maleate, atropine sulphate and diphenhydramine HCl) as separate compounds as well as in pharmaceutical preparations through the formation of their ion-pair (reineckate complexes) is described.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service