Skip to Content
Merck
All Photos(4)

Key Documents

P3289

Sigma-Aldrich

Potassium hexacyanoferrate(II) trihydrate

ACS reagent, 98.5-102.0%

Synonym(s):

Potassium ferrocyanide, Yellow prussiate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
K4Fe(CN)6 · 3H2O
CAS Number:
Molecular Weight:
422.39
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21

grade

ACS reagent

Assay

98.5-102.0%

form

powder

reaction suitability

reagent type: catalyst
core: iron

impurities

≤0.005% Insoluble matter

pH

8-10 (25 °C, 211 g/L)

mp

70 °C (lit.)

anion traces

chloride (Cl-): ≤0.01%
sulfate (SO42-): passes test (limit about 0.01%)

SMILES string

O.O.O.[K+].[K+].[K+].[K+].N#C[Fe-4](C#N)(C#N)(C#N)(C#N)C#N

InChI

1S/6CN.Fe.4K.3H2O/c6*1-2;;;;;;;;/h;;;;;;;;;;;3*1H2/q;;;;;;-4;4*+1;;;

InChI key

NRYOVBQTVIWDNU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Potassium hexacyanoferrate(II) trihydrate, commonly known as potassium ferrocyanide, is a commonly used anticaking agent. Its cytotoxic and genotoxic effect has been evaluated. The growth of chemical garden in aqueous potassium hexacyanoferrate solution droplets on a superhydrophobic surface has been reported. The inhibition efficiency of potassium ferrocyanide in phosphoric acid solution against the corrosion of mild steel has been investigated.

Application

Potassium hexacyanoferrate(II) trihydrate may be used in the following processes:
  • Preparation of Carrez I solution.
  • Milk sample preparation for gold nanoparticles (AuNPs) based colorimetric detection of melamine.
  • Preparation of ferrocyanide/ferricyanide reaction, a redox pair solution for electrochemical testing.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Factors influencing acrylamide formation in rye, wheat and spelt breads.
Przygodzka M, et al.
Journal of Cereal Science, 65, 96-102 (2015)
Studies on the inhibitive effect of potassium ferrocyanide on the corrosion of steel in phosphoric acid.
Hmamou DB, et al.
Research on Chemical Intermediates, 39(8), 3475-3485 (2013)
Mixing with herringbone-inspired microstructures: overcoming the diffusion limit in co-laminar microfluidic devices.
Marschewski J, et al.
Lab on a chip, 15(8), 1923-1933 (2015)
Pierre Osteil et al.
Development (Cambridge, England), 146(7) (2019-03-21)
During embryogenesis, the stringent regulation of Wnt activity is crucial for the morphogenesis of the head and brain. The loss of function of the Wnt inhibitor Dkk1 results in elevated Wnt activity, loss of ectoderm lineage attributes from the anterior
Thorsten Litzenburger et al.
Clinical science (London, England : 1979), 134(7), 921-939 (2020-04-03)
Maternal obesity determines obesity and metabolic diseases in the offspring. The white adipose tissue (WAT) orchestrates metabolic pathways, and its dysfunction contributes to metabolic disorders in a sex-dependent manner. Here, we tested if sex differences influence the molecular mechanisms of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service