Skip to Content
Merck
All Photos(3)

Documents

M3634

Sigma-Aldrich

Manganese(II) chloride tetrahydrate

ReagentPlus®, ≥99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
MnCl2 · 4H2O
CAS Number:
Molecular Weight:
197.91
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21

Quality Level

product line

ReagentPlus®

Assay

≥99%

form

powder

pH

3.5-6 (25 °C, 50 g/L)

mp

58 °C (lit.)

SMILES string

Cl[Mn]Cl.[H]O[H].[H]O[H].[H]O[H].[H]O[H]

InChI

1S/2ClH.Mn.4H2O/h2*1H;;4*1H2/q;;+2;;;;/p-2

InChI key

CNFDGXZLMLFIJV-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Manganese(II) chloride tetrahydrate is the tetrahydrate form of manganese(II) chloride. It is used in the preparation of manganese-based hybrid compounds.

Application

Manganese(II) chloride tetrahydrate can be used:      
  • As a promoter to synthesize dihydropyrimidin-2(1H)-ones by the condensation of aldehyde, β-dicarbonyl compounds, and urea under microwave conditions.      
  • As a catalyst to synthesize dihydro-2-oxypyrrole derivatives via one-pot four-component domino condensation of dialkyl acetylenedicarboxylate, formaldehyde, and amines.     
  • To synthesize active manganese oxide with potential application as an oxidant in organic synthesis.

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Dam. 1 - STOT RE 2

Target Organs

Brain

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 2

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Seong-Uk Jin et al.
Magnetic resonance imaging, 31(7), 1143-1149 (2013-05-11)
The cochlear plays a vital role in the sense and sensitivity of hearing; however, there is currently a lack of knowledge regarding the relationships between mechanical transduction of sound at different intensities and frequencies in the cochlear and the neurochemical
Lisha Luo et al.
NMR in biomedicine, 25(12), 1360-1368 (2012-05-11)
The aim of this study was to provide data on the dose dependence of manganese-enhanced MRI (MEMRI) in the visual pathway of experimental rats and to study the toxicity of MnCl₂ to the retina. Sprague-Dawley rats were intravitreally injected with
Geoffrey J Topping et al.
Medical physics, 40(4), 042502-042502 (2013-04-06)
Manganese(II) is employed as a contrast agent with magnetic resonance imaging (MRI) for study of neuronal activation in rats and mice. However, at the concentrations required for MRI, Mn may induce pharmacological or toxic effects. Positron emission tomography (PET) imaging
Tameshwar Ganesh et al.
Journal of magnetic resonance imaging : JMRI, 41(3), 806-813 (2014-03-05)
To investigate the potential of manganese (Mn)-enhanced MRI for sensitive detection and delineation of tumors that demonstrate little enhancement on Gd-DTPA. Eighteen nude rats bearing 1 to 2 cm in diameter orthotopic breast tumors (ZR75 and LM2) were imaged on
Marta Sidoryk-Wegrzynowicz et al.
Journal of neurochemistry, 122(4), 856-867 (2012-06-20)
Manganese (Mn) has been implicated in the impairment of the glutamate-glutamine cycling (GGC) by deregulation of Glu and glutamine (Gln) turnover in astrocytes. Here, we have examined possible mechanisms involved in the Mn(II)-mediated disruption of Glu turnover, including those related

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service