Skip to Content
Merck
All Photos(3)

Key Documents

C85727

Sigma-Aldrich

m-Cresol

99%

Synonym(s):

1-Hydroxy-3-methylbenzene, 3-Hydroxytoluene, 3-Methylphenol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H4OH
CAS Number:
Molecular Weight:
108.14
Beilstein:
506719
EC Number:
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.21
bp:
203 °C (lit.)
vapor pressure:
<1 mmHg ( 20 °C)

Agency

suitable for SM 2310
suitable for SM 2320

vapor density

3.72 (vs air)

vapor pressure

<1 mmHg ( 20 °C)

Assay

99%

form

liquid

autoignition temp.

1036 °F

expl. lim.

1.06-1.35 %, 150 °F

refractive index

n20/D 1.541 (lit.)

bp

203 °C (lit.)

mp

8-10 °C (lit.)

density

1.034 g/mL at 25 °C (lit.)

SMILES string

Cc1cccc(O)c1

InChI

1S/C7H8O/c1-6-3-2-4-7(8)5-6/h2-5,8H,1H3

InChI key

RLSSMJSEOOYNOY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

m-Cresol can be used as a solvent to synthesize:
  • Porous porphyrin-based polyimides for selective CO2 capture.
  • Thermoplastic polymers via thiol-ene Click reactions.

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

186.8 °F - closed cup

Flash Point(C)

86 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Synthesis of a polyimide porous porphyrin polymer for selective CO2 capture
Neti V, et al.
Journal of Chemistry (2015)
Segmented thermoplastic polymers synthesized by thiol-ene click chemistry: examples of thiol-norbornene and thiol-maleimide click reactions
Jin K, et al.
Macromolecules, 51, 3620-3631 (2018)
Min Ji Lee et al.
Materials (Basel, Switzerland), 13(6) (2020-03-18)
Fluidized beds have been utilized for various chemical and physical applications including heat transfer such as the gas-solid heat exchanger. It is advantageous to use carbon nanotubes (CNTs) with high thermal conductivity as bed materials for heat transfer enhancement in
Magdalena Kwiatkowska et al.
Polymers, 12(2) (2020-02-08)
In this work the preparation of polyamide 12 (PA12) based composites reinforced with pristine and surface-modified carbon nanotubes is reported. A qualitative and quantitative evaluation of multi-walled carbon nanotube functionalization with oxygen containing reactive groups achieved by different procedures of
Laurence A Angel et al.
The journal of physical chemistry. A, 110(35), 10392-10403 (2006-09-01)
Energy-resolved, competitive threshold collision-induced dissociation (TCID) methods are used to measure the gas-phase acidities of phenol, 3-methylphenol, 2,4,6-trimethylphenol, and ethanoic acid relative to hydrogen cyanide, hydrogen sulfide, and the hydroperoxyl radical using guided ion beam tandem mass spectrometry. The gas-phase

Related Content

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service