Skip to Content
Merck
All Photos(4)

Documents

B0394

Sigma-Aldrich

Boric acid

ACS reagent, ≥99.5%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H3BO3
CAS Number:
Molecular Weight:
61.83
EC Number:
MDL number:
UNSPSC Code:
12352106
eCl@ss:
38120104
PubChem Substance ID:
NACRES:
NA.21

grade

ACS reagent

Quality Level

Agency

suitable for SM 4500 - NH3
suitable for SM 5210

vapor pressure

2.6 mmHg ( 20 °C)

Assay

≥99.5%

form

powder

technique(s)

PCR: suitable

impurities

≤0.005% Insoluble in methanol
≤0.05% Nonvolatile with methanol

mp

160 °C (dec.) (lit.)

solubility

water: soluble

density

1.440 g/cm3

anion traces

chloride (Cl-): ≤0.001%
phosphate (PO43-): ≤0.001%
sulfate (SO42-): ≤0.01%

cation traces

Ca: ≤0.005%
Fe: ≤0.001%
heavy metals: ≤0.001% (by ICP)

SMILES string

OB(O)O

InChI

1S/BH3O3/c2-1(3)4/h2-4H

InChI key

KGBXLFKZBHKPEV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Boric acid is a weak monobasic Lewis acid(electron pair acceptor), which finds application in the areas of glass making, fire retardant, lubrication agent, buffer, and neutron absorber.

Application

Boric acid can be used as a catalyst to synthesize:
  • N-substituted cinnamamides via amidation reaction between cinnamic acid and benzylamines.
  • β-Aminoalcohols through the regioselective epoxides ring opening with aromatic amines in the presence of glycerol as a co-catalyst.
  • Organosulfur compounds via thia-Michael addition of thiols to α, β-unsaturated compounds.
It can also be used as an activator for the solvent-free reduction of aldehydes and ketones to alcohols in the presence of sodium borohydride.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1B

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Debin Wang et al.
ACS nano, 8(8), 7896-7904 (2014-07-16)
Building plasmonic nanostructures using biomolecules as scaffolds has shown great potential for attaining tunable light absorption and emission via precise spatial organization of optical species and antennae. Here we report bottom-up assembly of hierarchical plasmonic nanostructures using DNA origami templates
Jing Li et al.
American journal of physiology. Cell physiology, 306(10), C943-C960 (2014-03-22)
The human solute carrier (SLC26) family of anion transporters consists of 10 members (SLCA1-11, SLCA10 being a pseudogene) that encode membrane proteins containing ~12 transmembrane (TM) segments with putative N-glycosylation sites (-NXS/T-) in extracellular loops and a COOH-terminal cytosolic STAS
Yona Goldshmit et al.
Oncotarget, 5(18), 8602-8613 (2014-09-28)
The three oncogenes, ErbB receptors, Ras proteins and nucleolin may contribute to malignant transformation. Previously, we demonstrated that nucleolin could bind both Ras protein and ErbB receptors. We also showed that the crosstalk between the three proteins facilitates anchorage independent
Juliane Schott et al.
Dalton transactions (Cambridge, England : 2003), 43(30), 11516-11528 (2014-05-23)
In the presence of polyborates (detected by (11)B-NMR) the formation of a weak Eu(III) borate complex (lg β11 ~ 2, estimated) was observed by time-resolved laser-induced fluorescence spectroscopy (TRLFS). This complex is a precursor for the formation of a solid
Hye Ryeo Lee et al.
Journal of chromatography. A, 1346, 117-122 (2014-05-09)
In liquid phase microextraction, high enrichment factors can be obtained using an acceptor phase of small volume. By hanging an acceptor drop at the separation capillary tip, single drop microextraction (SDME) can be in-line coupled with capillary electrophoresis (CE). The

Articles

Information on Isoelectric Focusing including what it is and how it is used. In order to ensure the high performance of analysis, isoelectric point (pI) standards are needed.

Acid and base chart lists the strength of acids and bases (strongest to weakest) in order. Simple to use laboratory reference chart for scientists, researchers and lab technicians.

Acid and base chart lists the strength of acids and bases (strongest to weakest) in order. Simple to use laboratory reference chart for scientists, researchers and lab technicians.

Acid and base chart lists the strength of acids and bases (strongest to weakest) in order. Simple to use laboratory reference chart for scientists, researchers and lab technicians.

See All

Related Content

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service