Skip to Content
Merck
All Photos(1)

Documents

71840

Sigma-Aldrich

Sodium perborate tetrahydrate

purum p.a., 96% (RT)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaBO3 · 4H2O
CAS Number:
Molecular Weight:
153.86
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21

grade

purum p.a.

Quality Level

Assay

96% (RT)

form

powder or crystals

concentration

≥10% (active oxygen)

pH

10-10.4 (25 °C, 10 g/L)

mp

60 °C (dec.) (lit.)

anion traces

chloride (Cl-): ≤500 mg/kg
sulfate (SO42-): ≤6000 mg/kg

cation traces

Ca: ≤200 mg/kg
Cd: ≤50 mg/kg
Co: ≤50 mg/kg
Cu: ≤50 mg/kg
Fe: ≤50 mg/kg
K: ≤100 mg/kg
Ni: ≤50 mg/kg
Pb: ≤50 mg/kg
Zn: ≤50 mg/kg

SMILES string

O.O.O.O.[Na+].[O-]B1OO1

InChI

1S/BO3.Na.4H2O/c2-1-3-4-1;;;;;/h;;4*1H2/q-1;+1;;;;

InChI key

IBDSNZLUHYKHQP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Sodium perborate tetrahydrate is a hydrated sodium salt. It participates in various oxidation reactions of anilines and sulfides.

Application

Sodium perborate tetrahydrate may be employed for the following studies:
  • Transformation of iodoarenes to (diacetoxyiodo)arenes.
  • Oxidation of Π-deficient azines to afford N-oxides.
  • Oxidation of sulfur containing heterocyclic compounds to afford S,S-dioxides.
  • Microwave-assisted oxidative hydration of various aromatic and aliphatic nitriles.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Repr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Microwave-promoted transformation of nitriles to amides with aqueous sodium perborate.
Sharifi A, et al.
Synthetic Communications, 31(3), 431-434 (2001)
Further functional group oxidations using sodium perborate.
McKillop A and Kemp D.
Tetrahedron, 45(11), 3299-3306 (1989)
Functional group oxidation using sodium perborate1.
McKillop A and Tarbin JA.
Tetrahedron, 43(8), 1753-1758 (1987)
Eduardo Buozi Moffa et al.
Journal of dentistry, 39 Suppl 3, e65-e71 (2011-11-03)
This randomised clinical study evaluated the effect of chemical disinfection with sodium perborate or chlorhexidine on the colour stability of a hard chairside reline resin during six months. Hard chairside reline resin (Tokuyama Rebase Fast II) was used to reline
Milton Carlos Kuga et al.
Dental traumatology : official publication of International Association for Dental Traumatology, 28(3), 238-242 (2011-11-22)
To compare the fracture resistance of bovine teeth after intracoronal bleaching with sodium percarbonate (SPC) or sodium perborate (SP) mixed with water or 20% hydrogen peroxide (HP). Fifty extracted bovine teeth were divided into four experimental groups (G1-G4) and one

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service