Skip to Content
Merck
All Photos(1)

Key Documents

676934

Sigma-Aldrich

1,4-Dioxane

ACS reagent, ≥99.0%, contains <=25 ppm BHT as stabilizer

Synonym(s):

Diethylene oxide, Dioxane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H8O2
CAS Number:
Molecular Weight:
88.11
Beilstein:
102551
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.21
grade:
ACS reagent
technique(s):
thin layer chromatography (TLC): suitable
bp:
100-102 °C (lit.)
vapor pressure:
27 mmHg ( 20 °C)
40 mmHg ( 25 °C)

grade

ACS reagent

vapor density

3 (vs air)

vapor pressure

27 mmHg ( 20 °C)
40 mmHg ( 25 °C)

Assay

≥99.0%

form

liquid

autoignition temp.

356 °F

expl. lim.

22 %

technique(s)

thin layer chromatography (TLC): suitable

impurities

≤0.0016 meq/g Titr. acid
≤0.005% Peroxide as (H2O2)
≤0.01% Carbonyl (as HCHO)
≤0.05% water

evapn. residue

≤0.005%

color

APHA: ≤20

refractive index

n20/D 1.422 (lit.)

pH

6.0-8 (20 °C, 500 g/L)

bp

100-102 °C (lit.)

mp

10-12 °C (lit.)

transition temp

freezing point ≥11.0

density

1.034 g/mL at 25 °C (lit.)

SMILES string

C1COCCO1

InChI

1S/C4H8O2/c1-2-6-4-3-5-1/h1-4H2

InChI key

RYHBNJHYFVUHQT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1,4-Dioxane is widely used as a solvent for various organic compounds in industry. It is a non-biodegradable contaminant found in industrial effluents. Its oxidative degradation by using oxygen and ozone has been useful in converting it into biodegradable products, which can be easily removed by conventional methods.

Application

1,4-Dioxane may be employed as solvent in the synthesis of N-(4-(prop-2-ynyloxy)phenyl)acetamide and 3-(4-(prop-2-ynyloxy)phenyl)-3,4-dihydro-2H-benzo[e][1,3]oxazine.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1B - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

51.8 °F - closed cup

Flash Point(C)

11 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Self-curable benzoxazine functional polybutadienes synthesized by click chemistry.
Kukut M, et al.
Designed Monomers and Polymers, 12(2), 167-176 (2009)
Oxidation and biodegradability enhancement of 1,4-dioxane using hydrogen peroxide and ozone.
C D Adams et al.
Environmental science & technology, 28(11), 1812-1818 (1994-10-01)
Xiaohua Liu et al.
Biomaterials, 31(2), 259-269 (2009-09-29)
It remains a challenge to synthesize functional materials that can develop advanced scaffolding architectures for tissue engineering. In this study, a series of biodegradable amphiphilic poly(hydroxyalkyl (meth)acrylate)-graft-poly(l-lactic acid) (PHAA-g-PLLA) copolymers have been synthesized and fabricated into nano-fibrous scaffolds. These copolymers
Kiyoaki Ishimoto et al.
Biomacromolecules, 13(11), 3757-3768 (2012-09-19)
For developing broader application of biobased polymers, graft copolymers and comb polymers having poly(lactic acid) (PLA) side chains have been synthesized by using a macromonomer technique. PLA macromonomers (MMm) having a methacryloyl polymerizable group with different PLA chain length with
Alkene trifluoromethylation coupled with C-C bond formation: construction of trifluoromethylated carbocycles and heterocycles.
Hiromichi Egami et al.
Angewandte Chemie (International ed. in English), 52(14), 4000-4003 (2013-02-27)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service